Skip to Content
Merck

Skip To

C9426

CHAPS hydrate

BioReagent, suitable for electrophoresis, ≥98% (HPLC)

Synonym(s):

3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, CHAPS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C32H58N2O7S · xH2O
CAS Number:
Molecular Weight:
614.88 (anhydrous basis)
NACRES:
NB.22
PubChem Substance ID:
UNSPSC Code:
12161902
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

zwitterionic

Quality Segment

product line

BioReagent

assay

≥98% (HPLC)

form

powder

mol wt

614.88 g/mol (anhydrous basis)

technique(s)

electrophoresis: suitable, protein quantification: suitable

impurities

<0.04% DMF

mp

157 °C (dec.) (lit.)

solubility

water: 50 mg/mL, clear, colorless

suitability

suitable for electrophoresis

storage temp.

room temp

SMILES string

O.C[C@H](CCC(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C32H58N2O7S.H2O/c1-21(8-11-29(38)33-14-6-15-34(4,5)16-7-17-42(39,40)41)24-9-10-25-30-26(20-28(37)32(24,25)3)31(2)13-12-23(35)18-22(31)19-27(30)36;/h21-28,30,35-37H,6-20H2,1-5H3,(H-,33,38,39,40,41);1H2/t21-,22+,23-,24-,25+,26+,27-,28+,30+,31+,32-;/m1./s1

InChI key

SJCUTFKCLFLIFE-JWTJKVBLSA-N

General description

CHAPS is a non-denaturing, zwitterionic sulfobetaine detergent. It is mild and solubilizes membrane proteins and receptors. It is useful for breaking protein-protein interactions. It is a derivative of the naturally occurring bile salts. It has small micellar molecular weight (6,150) and high critical micellar concentration (6 - 10 mM) allowing it to be removed from samples by dialysis.

Application

CHAPS has been used:
  • For membrane permeabilization of cells
  • As a component in lysis buffer for homogenization of liver tissues as well as adipose tissue
  • As a component in denaturing buffer in enzyme characterization
  • As a component of 2D- sample buffer in 2D electrophoresis
  • For protein solubilization in isoelectric focusing

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C3023850500PC5070
technique(s)

electrophoresis: suitable, protein quantification: suitable

technique(s)

dialysis: suitable, electrophoresis: suitable

technique(s)

protein quantification: suitable

technique(s)

protein purification: suitable, protein quantification: suitable

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

>99% (TLC)

assay

≥98% (TLC)

Quality Level

200

Quality Level

400

Quality Level

-

Quality Level

400

product line

BioReagent

product line

-

product line

-

product line

BioXtra

form

powder

form

powder

form

powder

form

powder

solubility

water: 50 mg/mL, clear, colorless

solubility

water: 50 mg/mL

solubility

-

solubility

H2O: 0.1 M, clear, colorless


Still not finding the right product?

Explore all of our products under CHAPS hydrate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How do I store CHAPS, once it's dissolved in H2O?

    1 answer
    1. Solutions prepared at 100 mM in water are stable for at least 1 year when stored refrigerated. Note that a prepared solution is available. Please see the link below to review:
      https://www.sigmaaldrich.com/product/sigma/19899

      Helpful?

Reviews

No rating value

Active Filters