Skip to Content
Merck
All Photos(1)

Key Documents

C9393

Sigma-Aldrich

Cloxacillin sodium salt monohydrate

β-lactamase-resistant antibiotic

Synonym(s):

Cloxacillin, Sodium cloxacillin monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C19H17ClN3O5SNa · H2O
CAS Number:
Molecular Weight:
475.88
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

form

powder or crystals

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

O.[Na+].Cc1onc(-c2ccccc2Cl)c1C(=O)N[C@H]3[C@H]4SC(C)(C)[C@@H](N4C3=O)C([O-])=O

InChI

1S/C19H18ClN3O5S.Na.H2O/c1-8-11(12(22-28-8)9-6-4-5-7-10(9)20)15(24)21-13-16(25)23-14(18(26)27)19(2,3)29-17(13)23;;/h4-7,13-14,17H,1-3H3,(H,21,24)(H,26,27);;1H2/q;+1;/p-1/t13-,14+,17-;;/m1../s1

InChI key

KCUWTKOTPIUBRI-VICXVTCVSA-M

General description

Chemical structure: ß-lactam

Application

Cloxacillin is effective against infections caused by penicillin G-resistant staphylococci. It was used in studies of topical antibacterial agents for burn and crush wounds.

Biochem/physiol Actions

Cloxacillin is a semi-synthetic antibiotic and a chlorinated derivative of oxacillin. It inhibits the last stage of bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs), which results in cell lysis. Cell lysis is mediated by bacterial cell wall autolytic enzymes. Cloxacillin is often used as AmpC β-lactamase inhibitor in combination with β-lactam antibiotics.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A New Antibacterial Agent: Sodium Cloxacillin Monohydrate (Tegopen)
Journal of the American Medical Association, 195, 569-570 (1966)
Methenamine salts as topical antibacterial agent. I. Experimental studies for burn and crush wound.
T Matsumoto et al.
Archives of surgery (Chicago, Ill. : 1960), 101(1), 71-77 (1970-07-01)
Anna Stefánsdóttir et al.
Acta orthopaedica, 84(1), 87-91 (2013-02-16)
Prosthetic joint infections can be caused by bacteria derived from the patient's skin. The aim of the study was: (1) to determine which bacteria colonize the nose and groin in patients planned for primary hip or knee arthroplasty, (2) to
Srecko Marusic et al.
International journal of clinical pharmacology and therapeutics, 50(6), 431-433 (2012-06-09)
To present the case of warfarin-cloxacillin interaction that resulted in an increased international normalized ratio (INR). A 70-year-old man had been treated with warfarin for atrial fibrillation. He was hospitalized because of superficial thrombophlebitis of the left median cubital vein
Hossein Khalili et al.
European journal of clinical pharmacology, 69(3), 721-724 (2012-09-05)
Three patients were admitted to the Imam Hospital, Tehran, Iran with a diagnosis of bacterial endocarditis. The patients had indications for valve replacement surgery and anticoagulant therapy. The administration of cloxacillin reduced the effect of warfarin, and subsequent increases in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service