Skip to Content
Merck
All Photos(1)

Key Documents

C9373

Sigma-Aldrich

CP-96345

≥98% (HPLC)

Synonym(s):

(2S,3S)-cis-2-(Diphenylmethyl)-N-[(2-methoxyphenyl)methyl]-1-azabicyclo[2.2.2]octan-3-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32N2O
CAS Number:
Molecular Weight:
412.57
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: 24 mg/mL

storage temp.

room temp

SMILES string

COC1=CC=CC=C1CN[C@@H]2[C@H](C(C3=CC=CC=C3)C4=CC=CC=C4)N5CCC2CC5

InChI

1S/C28H32N2O/c1-31-25-15-9-8-14-24(25)20-29-27-23-16-18-30(19-17-23)28(27)26(21-10-4-2-5-11-21)22-12-6-3-7-13-22/h2-15,23,26-29H,16-20H2,1H3/t27-,28-/m0/s1

InChI key

FLNYLINBEZROPL-NSOVKSMOSA-N

Biochem/physiol Actions

CP-96345 is a selective NK1 antagonist. CP96345 inhibits substance P-induced salivation in the rat by classical in vivo bioassay, but did not inhibit NK2, NK3, or numerous other receptors.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dwight L Evans et al.
Biological psychiatry, 63(9), 899-905 (2007-10-20)
Natural killer (NK) cells play an important role in innate immunity and are involved in the host defense against human immunodeficiency virus (HIV) infection. This study examines the potential role of three underlying regulatory systems that have been under investigation
Luc Jasmin et al.
Vascular pharmacology, 45(4), 243-250 (2006-08-11)
To determine whether noradrenaline (NA) is an essential neurotransmitter for addictive and appetitive behaviors, we measured drug and food seeking in transgenic mice lacking dopamine beta-hydroxylase (Dbh), the enzyme responsible for synthesizing NA. Using the conditioned place preference test (CPP)
Stefan Nessler et al.
Journal of neuroimmunology, 179(1-2), 1-8 (2006-08-15)
Substance P (SP) is an excitatory neurotransmitter in the central and peripheral nervous system. Most of its physiological functions are mediated through binding to the neurokinin-1 receptor (NK-1R). Recently, proinflammatory properties of SP have been described. In this study we
Nasser Haddjeri et al.
European journal of pharmacology, 600(1-3), 64-70 (2008-10-22)
Substance P (neurokinin-1; NK1) receptor antagonists represent a putative new class of antidepressant/anxiolytic drugs. Using in vivo electrophysiological paradigms in rats, this study examined the effects of acute, sub-acute and long-term administration of these drugs on the firing of rat
Xu Wang et al.
The American journal of pathology, 169(5), 1663-1670 (2006-10-31)
Opioid withdrawal is a crucial and recurring event during the course of opioid abuse that has a negative impact on the immune system. In this study, we investigated whether abrupt withdrawal (AW) or precipitated withdrawal (PW) potentiates human immunodeficiency virus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service