Skip to Content
Merck

Skip To

C9249

CryoSOfree DMSO-free Cryopreservation Medium

greener alternative

Synonym(s):

cell freezing medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.75
UNSPSC Code:
12352207
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

cell culture | mammalian: suitable, cryopreservation: suitable

greener alternative category

shipped in

wet ice

storage temp.

2-8°C

General description

Cultured cells in a suspension can be cryopreserved with minimal damage from freezing and thawing steps. CryoSOfree DMSO-free cryopreservation medium preserves them with high viability and physiological activity. This medium is recommended for the preservation of stem cells, hepatocytes, tissue sample and a wide variety of cell types and established cell lines. CryoSOfree DMSO-free cryopreservation medium is a proprietary formulation, containing no DMSO nor animal proteins. It is ready-to-use and complete with no additives required.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of DMSO for cryopreservation of cell cultures. For more information see here.

Application

CryoSOfree is a chemically defined DMSO-free cryopreservation medium. It is animal component free and has been shown[1][2] to preserve the potential of stem cell for differentiation.

Features and Benefits

  • Preserves pluripotency of stem cells
  • Low cytotoxicity
  • Chemically defined, serum free, protein free
  • Ready to use


The following cell lines have been recovered at ≥90% following cryopreservation with CryoSOfree:
L929, KB, MC3T3, B16F1, Colon26, CaCO2, HT1080, MG63

Preparation Note

1 ml of CyroSOfree is sufficient for 5x105- 5x106 cells.

Legal Information

CryoSOfree is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C6295C6164C2999
technique(s)

cell culture | mammalian: suitable, cryopreservation: suitable

technique(s)

cell culture | mammalian: suitable, cryopreservation: suitable

technique(s)

cell culture | mammalian: suitable, cryopreservation: suitable

technique(s)

cell culture | mammalian: suitable, cryopreservation: suitable

form

liquid

form

liquid

form

liquid

form

liquid

shipped in

wet ice

shipped in

dry ice

shipped in

dry ice

shipped in

ambient

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

-

greener alternative product characteristics

-

greener alternative product characteristics

-


pictograms

Corrosion

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters