Skip to Content
Merck
All Photos(1)

Documents

C9140

Sigma-Aldrich

Convallatoxin

≥65%

Synonym(s):

3β,5α,14-Trihydroxy-19-oxo-5β,20(22)-cardenolide 3-(6-deoxy-α-L-mannopyranoside), Strophanthidin α-L-rhamnopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H42O10
CAS Number:
Molecular Weight:
550.64
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

concentration

≥65%

SMILES string

C[C@@H]1O[C@@H](O[C@H]2CC[C@]3(C=O)[C@H]4CC[C@]5(C)[C@H](CC[C@]5(O)[C@@H]4CC[C@]3(O)C2)C6=CC(=O)OC6)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C29H42O10/c1-15-22(32)23(33)24(34)25(38-15)39-17-3-8-27(14-30)19-4-7-26(2)18(16-11-21(31)37-13-16)6-10-29(26,36)20(19)5-9-28(27,35)12-17/h11,14-15,17-20,22-25,32-36H,3-10,12-13H2,1-2H3/t15-,17-,18+,19-,20+,22-,23+,24+,25-,26+,27-,28-,29-/m0/s1

InChI key

HULMNSIAKWANQO-JQKSAQOKSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A cardiac glycoside found in lily-of-the-valley with digitalis-like cardiac effects.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Use of physicochemical analytical methods for assessing the interaction of various drug forms in therapeutic practice].
M N Tsarevskaia et al.
Farmakologiia i toksikologiia, 49(5), 42-44 (1986-09-01)
B M Anner et al.
The American journal of physiology, 262(5 Pt 2), F843-F848 (1992-05-01)
The investigation of active Na-K transport inhibition by mercury is difficult to perform in a cell because of the presence of numerous other membrane and intracellular proteins modifiable by mercury. Thus purified Na-K-adenosinetriphosphatase (ATPase) molecules performing active transport in an
[Effect of corglycon and vasoactive agents on systemic and cerebral hemodynamics in cerebral infarct].
S M Vinichuk
Vrachebnoe delo, (7)(7), 28-32 (1987-07-01)
[Prevention of frequent paroxysmal ventricular tachycardia by combined use of anti-arrhythmia drugs].
D F Kesler
Klinicheskaia meditsina, 59(11), 98-99 (1981-11-01)
[Effect of corglycon and cavinton on the hemodynamics of patients with initial manifestations of insufficiency of cerebral circulation].
M M Ishchenko et al.
Vrachebnoe delo, (5)(5), 53-55 (1988-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service