Skip to Content
Merck
All Photos(6)

Key Documents

C8546

Sigma-Aldrich

Cellulase from Trichoderma reesei ATCC 26921

greener alternative

lyophilized powder, ≥1 unit/mg solid

Synonym(s):

1,4-(1,3:1,4)-β-D-Glucan 4-glucano-hydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥1 unit/mg solid

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

solubility

deionized water: soluble 5.0 mg/mL (Sterile; In the presence of 0.15% polyhexamethylene biguanide (PHMB).)

greener alternative category

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in cellulosic ethanol research. For more information see the article in biofiles and Enzymes for Alternative Energy Research.

Application

Cellulase from Sigma has been used to degrade cello-oligosaccharides into glucose; to study the biodegradability of bioabsorbable bacterial cellulose (BBC) material.
Cellulase from Trichoderma reesei has been used in a study to assess the differential regulation of the cellulase transcription factors XYR1, ACE2, and ACE. Cellulase from Trichoderma reesei has also been used in a study to investigate the liquefaction of hydrothermally pretreated wheat straw at high-solids content.

Biochem/physiol Actions

Cellulases are enzymes that hydrolyze cellulose to glucose. Cellulase is used to study the development of occupational asthma in the detergent, pharmaceutical, baking, and enzyme production industries. Cellulase is added to detergents to improve cleansing properties. It is also applied in the biodegradation of paper products such as foolscap paper, filter paper, newspaper and office paper as well as microcrystalline cellulose.

Unit Definition

One unit will liberate 1.0 μmole of glucose from cellulose in one hr at pH 5.0 at 37 °C (2 hr incubation time).

Preparation Note

Dissolves in sterile deionized (DI) water in the presence of 0.15% polyhexamethylene biguanide (PHMB) at 5 mg/mL concentration.

substrate

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming-Li Yong et al.
Microscopy research and technique, 79(9), 838-844 (2016-07-01)
Rice false smut, caused by the fungal pathogen Villosiclava virens, is one of the most important rice diseases in the world. Previous studies reported that the pathogen has less number of cell wall-degraded genes and attacks dominantly rice stamen filaments
Thomas Portnoy et al.
Eukaryotic cell, 10(2), 262-271 (2010-12-21)
Due to its capacity to produce large amounts of cellulases, Trichoderma reesei is increasingly being investigated for second-generation biofuel production from lignocellulosic biomass. The induction mechanisms of T. reesei cellulases have been described recently, but the regulation of the genes
J P H van Wyk et al.
Bioresource technology, 86(1), 21-23 (2002-11-08)
Environmental issues such as the depletion of non-renewable energy resources and pollution are topical. The extent of solid waste production is of global concern and development of its bioenergy potential can combine issues such as pollution control and bioproduct development
Nóra Szijártó et al.
Bioresource technology, 102(2), 1968-1974 (2010-10-05)
Enzymatic liquefaction was studied by measuring continuously the flowability change of high-solids lignocellulose substrates using a real time viscometric method. Hydrolysis experiments of hydrothermally pretreated wheat straw were carried out with purified enzymes from Trichoderma reesei; Cel7A, Cel6A, Cel7B, Cel5A
Marko Riedel et al.
The Journal of eukaryotic microbiology, 56(2), 130-135 (2009-05-22)
Transgenic Phytophthora ramorum strains that produce green fluorescent protein (GFP) constitutively were obtained after stable DNA integration using a polyethylene glycol and CaCl₂-based transformation protocol. Green fluorescent protein production was studied in developing colonies and in different propagules of the

Protocols

To standardize an enzymatic assay procedure of cellulase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service