Skip to Content
Merck
All Photos(1)

Key Documents

C7869

Sigma-Aldrich

CEP-701 hydrate

≥98% (HPLC)

Synonym(s):

KT-5555 hydrate, Lestaurtinib hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H21N3O4 · xH2O
CAS Number:
Molecular Weight:
439.46 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: >10 mg/mL

storage temp.

−20°C

SMILES string

O.C[C@]12O[C@H](C[C@]1(O)CO)n3c4ccccc4c5c6C(=O)NCc6c7c8ccccc8n2c7c35

InChI

1S/C26H21N3O4.H2O/c1-25-26(32,12-30)10-18(33-25)28-16-8-4-2-6-13(16)20-21-15(11-27-24(21)31)19-14-7-3-5-9-17(14)29(25)23(19)22(20)28;/h2-9,18,30,32H,10-12H2,1H3,(H,27,31);1H2/t18-,25+,26+;/m1./s1

InChI key

LPOQWFBZBJTLEO-GMEYHWBASA-N

Application

CEP-701 hydrate has been used as a tyrosine kinase inhibitor:
  • to study its effects on early growth response protein (EGR) genes and nerve growth factor (NGF) stimulation in human epithelial cells
  • as a supplement in reservoir solution for co-crystallization studies with human receptor-interacting protein kinase 4 (RIPK4)
  • as an FMS-like tyrosine kinase 3 (FLT3) inhibitor-gold nanoparticle conjugate to study its effects on acute myeloid leukemia

Biochem/physiol Actions

CEP-701 hydrate, also known as Lestaurtinib, can repress Janus kinase 2/signal transducer and activator of transcription 5 (JAK2/STAT5) signaling by the specific inhibition of JAK2.
CEP-701 is a Flt-3 receptor tyrosine kinase inhibitor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bobe Petrushev et al.
International journal of nanomedicine, 11, 641-660 (2016-03-02)
Every year, in Europe, acute myeloid leukemia (AML) is diagnosed in thousands of adults. For most subtypes of AML, the backbone of treatment was introduced nearly 40 years ago as a combination of cytosine arabinoside with an anthracycline. This therapy
Tania Diaz et al.
PloS one, 6(4), e18856-e18856 (2011-05-03)
Standard cytotoxic chemotherapy for Hodgkin Lymphoma (HL) has changed little in 30 years; the treatment for patients with relapsed or refractory disease remains challenging and novel agents are under development. JAK/STAT constitutive activation plays an important role in the pathogenesis
Timea Simon et al.
Nanoscale research letters, 10(1), 466-466 (2015-12-03)
Releasing drug molecules at the targeted location could increase the clinical outcome of a large number of anti-tumor treatments which require low systemic damage and low side effects. Nano-carriers of drugs show great potential for such task due to their
Christine S Huang et al.
Structure (London, England : 1993), 26(5), 767-777 (2018-05-01)
Receptor-interacting protein kinase 4 (RIPK4) is a highly conserved regulator of epidermal differentiation. Members of the RIPK family possess a common kinase domain as well as unique accessory domains that likely dictate subcellular localization and substrate preferences. Mutations in human
M Rochman et al.
Mucosal immunology, 8(4), 785-798 (2014-11-13)
Although interleukin (IL)-13 and neurotrophins are functionally important for the pathogenesis of immune responses, the interaction of these pathways has not been explored. Herein, by interrogating IL-13-induced responses in human epithelial cells we show that neurotrophic tyrosine kinase receptor, type

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service