Skip to Content
Merck

Skip To

C7757

S-Carboxymethyl-L-cysteine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C5H9NO4S
CAS Number:
Molecular Weight:
179.19
UNSPSC Code:
12352209
NACRES:
NA.26
PubChem Substance ID:
EC Number:
211-327-5
Beilstein/REAXYS Number:
1725012
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

>98% (TLC)

form

powder

technique(s)

cell culture | mammalian: suitable

color

white

mp

200 °C

storage temp.

2-8°C

SMILES string

N[C@@H](CSCC(O)=O)C(O)=O

InChI

1S/C5H9NO4S/c6-3(5(9)10)1-11-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1

InChI key

GBFLZEXEOZUWRN-VKHMYHEASA-N

Biochem/physiol Actions

S-Carboxymethyl-L-cysteine is studied as a small molecule mucoactive drug in vivo. These studies include analyzing the oxidative metabolism of S-carboxymethyl-L-cysteine by enzymes such as phenylalanine monooxygenase(s).


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters