Skip to Content
Merck
All Photos(2)

Key Documents

C7464

Sigma-Aldrich

Monoclonal Anti-Cyclin D1 antibody produced in mouse

clone DCS-6, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

DCS-6, monoclonal

mol wt

antigen 36 kDa (several lower and higher molecular weight bands)

contains

15 mM sodium azide

species reactivity

rat, mouse, human, monkey

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunohistochemistry (frozen sections): suitable
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 1:200 using a human tumor cell line extract

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCND1(595)
mouse ... Ccnd1(12443)
rat ... Ccnd1(58919)

General description

Monoclonal Anti-Cyclin D1 (mouse IgG2a isotype) is derived from the DCS-6 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Cyclin D1 protein is a G1-phase cyclin in mammalian cells. This protein is a D-type cyclin. The CCD1 gene is located on the human chromosome at 11q13.3.

Specificity

Does not cross-react with other D-type cyclins.

Immunogen

recombinant human Cyclin D1 protein.

Application

Monoclonal Anti-Cyclin D1 antibody produced in mouse may be used in:
  • immunoblotting
  • immunofluorescence
  • immunocytochemistry
  • immunoprecipitation
  • immunohistochemistry
  • antibody-mediated knock-out experiments

Biochem/physiol Actions

Cyclin D1 is involved in regulating cyclin-dependent kinase (CDK) family members. Cyclin D1 and D2 are affiliated with CDK4 and/or CDK6 kinase activities that are observed in mid-G1 and culminate in the late G1 phase. Amplification of the cyclin D1 gene is observed in a significant percentage of cancers, including breast, bladder carcinoma, mantle cell lymphomas, head and neck cancers. This gene is also amplified in esophageal squamous cell carcinomas.

Physical form

The product is provided as ascites fluid with 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mu Yao et al.
The British journal of nutrition, 108(3), 424-430 (2011-11-10)
We have developed a blend of food extracts commonly consumed in the Mediterranean and East Asia, named blueberry punch (BBP), with the ultimate aim to formulate a chemoprevention strategy to inhibit prostate cancer progression in men on active surveillance protocol.
H Nakagawa et al.
Cancer, 76(4), 541-549 (1995-08-15)
Oncogene activation and tumor suppressor gene inactivation have been implicated in the genetic basis of esophageal squamous cell carcinoma (ESCC). Cyclin D1, an oncogene that has a critical role in G1 progression of the cell cycle, has been observed to
Takeshi Shimamura et al.
Cancer research, 65(14), 6401-6408 (2005-07-19)
Somatic mutations in the kinase domain of the epidermal growth factor receptor (EGFR), including L858R and exon 19 deletions, underlie responsiveness to gefitinib and erlotinib in non-small cell lung cancer (NSCLC). Acquired resistance to these tyrosine kinase inhibitors is in
Christopher T Capaldo et al.
Molecular biology of the cell, 22(10), 1677-1685 (2011-03-18)
Coordinated regulation of cell proliferation is vital for epithelial tissue homeostasis, and uncontrolled proliferation is a hallmark of carcinogenesis. A growing body of evidence indicates that epithelial tight junctions (TJs) play a role in these processes, although the mechanisms involved
Yanhong Yang et al.
Oncology letters, 15(4), 5280-5284 (2018-03-20)
Nimotuzumab, a humanized IgG1 monoclonal antibody against epidermal growth factor receptor (EGFR), increases radiosensitivity in lung cancer. Cisplatin is an effective antitumor agent in lung cancer. In the present study, the antitumor activity of nimotuzumab combined with cisplatin was investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service