Skip to Content
Merck
All Photos(1)

Key Documents

C6534

Sigma-Aldrich

Anti-Chicken Egg Albumin antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-Albumin, chicken egg

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

chicken

technique(s)

indirect ELISA: 1:80,000
quantitative precipitin assay: 3-5 mg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

chicken ... Alb(396197)

General description

Chicken/hen′s egg albumin, also known as ovalbumin (OVA), is the most abundant egg white protein. It is surrounded by B-cell epitopes, which are bound by specific IgE antibodies. In addition, it also contains CD4+ T cell epitopes. Ovalbumin is a monomeric protein with a molecular mass of 45000 Da.

Application

Anti-Chicken Egg Albumin antibody produced in rabbit has been used in:
  • quantitative precipitin assay
  • indirect enzyme linked immunoassay (ELISA)
  • development of nonimmune rabbit antiserum against chicken ovalbumin

Biochem/physiol Actions

Chicken/hen′s egg albumin or ovalbumin (OVA) is a major allergen found in egg white and is widely used in allergy research. It has been widely used to determine the characteristics of class II major histocompatibility complex (MHC)-peptide binding and T-cell activation.

Preparation Note

treated to remove lipoproteins

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison between ovalbumin and ovalbumin peptide 323-339 responses in allergic mice: humoral and cellular aspects
Sun L Z, et al.
Scandinavian Journal of Immunology, 71(7), 1192-1195 (2010)
Lara Bosco et al.
Insects, 11(12) (2020-12-10)
Following its first detection in North Italy in 2012, H. halys has become a serious threat in many crops, including hazelnut. The present study aimed at investigating dispersal capacity and behavior in relation to host plants of overwintered adults of
Angelino T Tromp et al.
Disease models & mechanisms, 13(9) (2020-05-31)
Bacterial pathogens have evolved to secrete strong anti-inflammatory proteins that target the immune system. It was long speculated whether these virulence factors could serve as therapeutics in diseases in which abnormal immune activation plays a role. We adopted the secreted
Melissa Y Tjota et al.
ImmunoHorizons, 1(6), 101-108 (2018-04-10)
Tissue infiltration by circulating monocytes is a critical step in the initiation and augmentation of type 2 inflammatory responses in the lungs. Our studies demonstrate that IL-33-/- mice have a defect in monocyte extravasation from the vasculature to the lung
Changes in the antigenic and immunoglobulin E--binding properties of hen's egg albumin with the combination of heat and gamma irradiation treatment
Kim MJ, et al.
Journal of Food Protection, 65(7), 1192-1195 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service