Skip to Content
Merck
All Photos(3)

Key Documents

C6512

Sigma-Aldrich

Cholesteryl hemisuccinate

Synonym(s):

3β-Hydroxy-5-cholestene 3-hemisuccinate, 5-Cholesten-3β-ol 3-hemisuccinate, Cholesteryl hydrogen succinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H50O4
CAS Number:
Molecular Weight:
486.73
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

liquid crystal

SMILES string

[H][C@@]([C@H](C)CCCC(C)C)([C@@]1(C)CC[C@@]23[H])CC[C@]1([C@](CC=C4C[C@H](CC[C@]34C)OC(CCC(O)=O)=O)2[H])[H]

InChI

1S/C31H50O4/c1-20(2)7-6-8-21(3)25-11-12-26-24-10-9-22-19-23(35-29(34)14-13-28(32)33)15-17-30(22,4)27(24)16-18-31(25,26)5/h9,20-21,23-27H,6-8,10-19H2,1-5H3,(H,32,33)/t21-,23+,24+,25-,26+,27+,30+,31-/m1/s1

InChI key

WLNARFZDISHUGS-MIXBDBMTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cholesteryl hemisuccinate (CHEMS) is an acidic cholesterol ester. It self assembles into bilayers in alkaline and neutral aqueous media. CHEMS exhibits pH sensitive polymorphism. It consists of succinic acid esterified to the beta-hydroxyl group of cholesterol. This chemical modification results in the ability of CHEMS to adopt a lamellar assembly in suitable aqueous media, whereas cholesterol forms monohydrate crystals in an aqueous environment.[1]

Application

  • Experimental determination and computational interpretation of biophysical properties of lipid bilayers enriched by Cholesteryl Hemisuccinate: Provides insights into the effects of Cholesteryl Hemisuccinate on phospholipid bilayers, contributing to a deeper understanding of its biophysical properties (W Kulig et al., 2015).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cholesteryl hemisuccinate exhibits pH sensitive polymorphic phase behavior.
Hafez IM and Cullis PR
Biochim. Biophys. Acta Gen. Subj., 1463(1), 107-114 (2000)
Denitsa Momekova et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 32(4-5), 308-317 (2007-10-12)
A major hurdle towards in vivo utilization of pH-sensitive liposomes is their prompt sequestration by reticuloendothelial system and hence short circulation time. Prolonged circulation of liposomes is usually achieved by incorporation of pegylated lipids, which have been frequently reported to
Julia Lehtinen et al.
Journal of controlled release : official journal of the Controlled Release Society, 131(2), 145-149 (2008-08-12)
Cationic polymers are efficient gene delivery vectors in in vitro conditions, but these carriers can fail in vivo due to interactions with extracellular polyanions, i.e. glycosaminoglycans (GAG). The aim of this study was to develop a stable gene delivery vector
Zhe Zhang et al.
eLife, 7 (2018-05-11)
Transient receptor potential melastatin 2 (TRPM2) is a Ca2+-permeable cation channel required for immune cell activation, insulin secretion, and body heat control. TRPM2 is activated by cytosolic Ca2+, phosphatidyl-inositol-4,5-bisphosphate and ADP ribose. Here, we present the ~3 Å resolution electron
Michael Zocher et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(50), E3463-E3472 (2012-11-16)
The steroid cholesterol is an essential component of eukaryotic membranes, and it functionally modulates membrane proteins, including G protein-coupled receptors. To reveal insight into how cholesterol modulates G protein-coupled receptors, we have used dynamic single-molecule force spectroscopy to quantify the

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service