Skip to Content
Merck
All Photos(3)

Key Documents

C6427

Sigma-Aldrich

Clindamycin 2-phosphate

aminoglycoside antibiotic

Synonym(s):

Clindamycin 2-dihydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34ClN2O8PS
CAS Number:
Molecular Weight:
504.96
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.76
Pricing and availability is not currently available.

Quality Level

form

solid

solubility

DMSO: 224 mg/mL at 25 °C

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

CCC[C@@H]1C[C@H](N(C)C1)C(=O)NC([C@H](C)Cl)C2O[C@H](SC)[C@H](OP(O)(O)=O)[C@@H](O)[C@H]2O

InChI

1S/C18H34ClN2O8PS/c1-5-6-10-7-11(21(3)8-10)17(24)20-12(9(2)19)15-13(22)14(23)16(18(28-15)31-4)29-30(25,26)27/h9-16,18,22-23H,5-8H2,1-4H3,(H,20,24)(H2,25,26,27)/t9-,10+,11-,12?,13+,14-,15?,16+,18+/m0/s1

InChI key

UFUVLHLTWXBHGZ-MWBQRTRKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Clindamycin 2-phosphate is an aminoglycoside antibiotic that has been used to study the cytoxicity of antibiotics on human cell lines [1], Bacterial protein synthesis and peptide translation, and the inhibition of human Tyrosyl-DNA Phosphodiesterase [2].

Biochem/physiol Actions

Clindamycin 2-phosphate is a pharmacological tyrosyl-DNA phosphodiesterase (Tdp1) inhibitor. Clindamycin 2-phosphate can repair DNA topoisomerase I-DNA covalent complexes by hydrolyzing the tyrosyl-DNA bond. It inhibits protein synthesis in bacteria by binding the 50s ribosomal subunit. [2]. Spectrum of Activity: Gram positive cocci and taxoplasma. Especially active against anaerobic bacteria.
Spectrum of Activity: Gram positive cocci and taxoplasma. Especially active against anaerobic bacteria.
Mode of Action: Inhibits protein synthesis in bacterial by binding the 50s ribosomal subunit.

Other Notes

Antibacterial and antiprotozoal antibiotic of the licosamide class.
Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Lact. - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiyong Liao et al.
Molecular pharmacology, 70(1), 366-372 (2006-04-19)
DNA topoisomerase I (Top1) is the target of camptothecin, and novel Top1 inhibitors are in development as anticancer agents. Top1 inhibitors damage DNA by trapping covalent complexes between the Top1 catalytic tyrosine and the 3'-end of the broken DNA. Tyrosyl-DNA
Diane Thiboutot et al.
Journal of the American Academy of Dermatology, 59(5), 792-800 (2008-09-23)
We sought to evaluate efficacy, safety, and tolerability of a combination of clindamycin phosphate 1.2% and benzoyl peroxide 2.5% (clindamycin-BPO 2.5%) aqueous gel in moderate to severe acne vulgaris. A total of 2813 patients, aged 12 years or older, were
M Badawy Abdel-Naser et al.
Expert opinion on pharmacotherapy, 9(16), 2931-2937 (2008-10-22)
Clindamycin phosphate 1.2% together with tretinoin 0.025% as a gel (CTG) is a topical formulation of a fixed and stable combination approved by the FDA for the treatment of acne vulgaris in patients 12 years of age or older. The
Lawrence F Eichenfield et al.
Pediatric dermatology, 26(3), 257-261 (2009-08-27)
Treatments for mild to moderately severe acne usually combine retinoid and antimicrobial therapy. Recently, the US FDA approved the combination of 1.2% clindamycin (CLIN) and 0.025% tretinoin (RA) in a novel gel formulation for the treatment of mild to moderate
Vuk Uskoković et al.
Journal of biomedical materials research. Part A, 101(5), 1427-1436 (2012-11-02)
Osteomyelitis has been traditionally treated by the combination of long-term antibiotic therapies and surgical removal of diseased tissue. The multifunctional material was developed in this study with the aim to improve this therapeutic approach by: (a) enabling locally delivered and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service