Skip to Content
Merck
All Photos(5)

Key Documents

C6417

Sigma-Aldrich

Monoclonal Anti-Cytokeratin Peptide 7 antibody produced in mouse

clone LDS-68, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

LDS-68, monoclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunohistochemistry (frozen sections): suitable
indirect immunofluorescence: 1:200 using formalin-fixed, paraffin-embedded sections of human and animal tissue
microarray: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT7(3855)

Looking for similar products? Visit Product Comparison Guide

General description

Keratin, type II cytoskeletal 7 is a protein encoded by the KRT7 gene in humans. It is also known as cytokeratin-7 (CK-7), keratin-7 (K7), sarcolectin (SCL) and K2C7. Keratin 7 is a type II keratin and is a member of the keratin gene family. It is found on chromosome 12. It is specifically expressed in the simple epithelia lining of the internal organs and in the gland ducts as well as blood vessels.
Monoclonal anti-Cytokeratin Peptide 7 (mouse IgG1 isotype) is derived from the hybridoma produced the fusion of mouse myeloma cells and splenocytes of an immunized mouse. Cytokeratins are the intermediate filaments of endocrine cells. These proteins belongs to the intermediate filament (10 nm) superfamily of cytoskeletal proteins. Cytokeratin 7 (CK 7/ KRT7) is a low molecular weight cytokeratin. It is present in epithelial tissues of lung, breast and ovary.

Specificity

The antibody reacts specifically with most simple epithelia but shows no reaction with epithelial cells in the intestine or in the stomach.

Immunogen

cytoskeletal preparation of the RT4 human bladder carcinoma cell line.

Application

Monoclonal Anti-Cytokeratin Peptide 7 antibody produced in mouse has been used in:
  • immunohistochemical staining
  • western blotting
  • immunohistochemistry
  • immunofluorescence.

Biochem/physiol Actions

Cytokeratins may play an important role in transhepatic transport and canalicular secretion. Cytokeratin 7 (CK 7/ KRT7) only stains bile ducts in normal liver, hence it is considered as “bile duct keratin”. CK 7 is widely used in surgical pathology. It acts as a specific marker for mammary and extramammary Paget′s disease.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshiyuki Kasai et al.
Regenerative therapy, 4, 71-77 (2016-03-24)
Autologous oral mucosal epithelial cell sheets have been used for treating epithelial defects such as cornea and esophagus. The cell source of patients' oral mucosal epithelial cell sheet should be examined in normality because it has individual difference. In this
Brush biopsy of human oral mucosal epithelial cells as a quality control of the cell source for fabrication of transplantable epithelial cell sheets for regenerative medicine
Kasai Y, et al.
Regenerative therapy, 4, 71-77 (2016)
Immunohistology of endocrine tumors
Diagnostic Immunohistochemistry (Masson Publishing), 291-339 (2010)
M Rosenberg et al.
Cytogenetics and cell genetics, 57(1), 33-38 (1991-01-01)
We have localized the genes which encode the human type II epidermal keratins K5, K6a, and K6b and the simple epithelial keratin K7 (KRT5, KRT6A, KRT6B, and KRT7, respectively) to chromosome 12 using Southern blot analysis of somatic cell hybrids.
Immortalized human corneal epithelial cells for ocular toxicity and inflammation studies.
Offord EA, et al.
Investigative Ophthalmology & Visual Science, 40(6), 1091-1101 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service