Skip to Content
Merck
All Photos(2)

Documents

C6277

Sigma-Aldrich

Congo Red

Dye content, ≥85%, certified by the Biological Stain Commission, powder

Synonym(s):

Congo Red 4B, Cosmos Red, Cotton Red B, Cotton Red C, Direct Red 28, Direct Red R, Direct Red Y

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H22N6Na2O6S2
CAS Number:
Molecular Weight:
696.66
Colour Index Number:
22120
Beilstein:
3894858
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Congo Red, BioXtra, certified by the Biological Stain Commission

grade

certified by the Biological Stain Commission

product line

BioXtra

form

powder

composition

Dye content, ≥85%

mp

>360 °C (lit.)

solubility

H2O: 10 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Nc1c(cc(c2ccccc12)S([O-])(=O)=O)\N=N\c3ccc(cc3)-c4ccc(cc4)\N=N\c5cc(c6ccccc6c5N)S([O-])(=O)=O

InChI

1S/C32H24N6O6S2.2Na/c33-31-25-7-3-1-5-23(25)29(45(39,40)41)17-27(31)37-35-21-13-9-19(10-14-21)20-11-15-22(16-12-20)36-38-28-18-30(46(42,43)44)24-6-2-4-8-26(24)32(28)34;;/h1-18H,33-34H2,(H,39,40,41)(H,42,43,44);;/q;2*+1/p-2/b37-35+,38-36+;;

InChI key

IQFVPQOLBLOTPF-HKXUKFGYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Congo red has been used for the staining of β amyloid peptides.

Biochem/physiol Actions

Congo red is a benzidine-based anionic diazo dye. It is a histological dye which binds to many amyloid proteins and is used for the quantification of amyloid β-peptide aggregation. Congo red also interacts with β-D-glucans, polysaccharides containing continuous β-(1→4)-linked D-glucopyranosyl units and some hemicellulosic galactoglucomannans.

Suitability

Certified for use as a counterstain for alum based hematoxylin and in the Highman or the Churukian modification of Puchtler′s amyloid stain.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Efficient imaging of amyloid deposits in Drosophila models of human amyloidoses.
Berg I, et al.
Nature Protocols, 5, 935-935 (2010)
Alejandro Llanos-Chea et al.
Journal of pediatric gastroenterology and nutrition, 68(4), 509-516 (2018-11-13)
Enteric bacterial pathogens cause diarrheal disease and mortality at significant rates throughout the world, particularly in children younger than 5 years. Our ability to combat bacterial pathogens has been hindered by antibiotic resistance, a lack of effective vaccines, and accurate
Long-term treatment of thalidomide ameliorates amyloid-like pathology through inhibition of ?-secretase in a mouse model of Alzheimer's disease.
He P, et al.
PLoS ONE, 8, e55091-e55091 (2013)
Apolipoprotein E associates with beta amyloid peptide of Alzheimer's disease to form novel monofibrils. Isoform apoE4 associates more efficiently than apoE3.
Sanan DA, et al.
The Journal of Clinical Investigation, 94, 860-860 (1994)
Quantifying amyloid beta-peptide (Abeta) aggregation using the Congo red-Abeta (CR-abeta) spectrophotometric assay.
Klunk WE, et al.
Analytical Biochemistry, 266, 66-66 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service