Skip to Content
Merck
All Photos(1)

Documents

C5865

Sigma-Aldrich

CD437

≥98% (HPLC), solid

Synonym(s):

6-[3-(1-Adamantyl)-4-hydroxyphenyl]-2-naphthalene carboxylic acid, AHPN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H26O3
CAS Number:
Molecular Weight:
398.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

impurities

~0.5 mol/mol water

color

yellow

mp

271.6-276 °C

solubility

DMSO: >10 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

OC(=O)c1ccc2cc(ccc2c1)-c3ccc(O)c(c3)C45CC6CC(CC(C6)C4)C5

InChI

1S/C27H26O3/c28-25-6-5-22(20-1-2-21-11-23(26(29)30)4-3-19(21)10-20)12-24(25)27-13-16-7-17(14-27)9-18(8-16)15-27/h1-6,10-12,16-18,28H,7-9,13-15H2,(H,29,30)

InChI key

LDGIHZJOIQSHPB-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

CD437 is a retinoic acid receptor (RAR)γ-selective agonist, γ-selective retinoid; potent inducer of apoptosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Tan et al.
Frontiers in microbiology, 10, 2301-2301 (2019-10-28)
Enterococcus faecalis (E. faecalis), a biofilm-forming pathogen, causes nosocomial infections. In recent years, drug resistance by enterococci has become increasingly severe due to widespread antibiotic abuse. Therefore, novel antibacterial agents are urgently needed. In this study, the synthetic retinoid compound
Almudena Bosch et al.
Breast cancer research : BCR, 14(4), R121-R121 (2012-08-28)
Retinoic acid signaling plays key roles in embryonic development and in maintaining the differentiated status of adult tissues. Recently, the nuclear retinoic acid receptor (RAR) isotypes α, β and γ were found to play specific functions in the expansion and
Liyue Zhang et al.
Journal of molecular signaling, 5, 12-12 (2010-08-14)
A variety of pathways target CDKI p21WAF1/CIP1 expression at transcriptional, post-transcriptional as well as translational levels. We previously found that cell growth suppressing retinoid CD437 enhanced expression of p21WAF1/CIP1 and DNA damage inducible GADD45 proteins in part by elevating their
Takaharu Asano et al.
Frontiers in cardiovascular medicine, 9, 842641-842641 (2022-04-12)
Conventional drug screening methods search for a limited number of small molecules that directly interact with the target protein. This process can be slow, cumbersome and has driven the need for developing new drug screening approaches to counter rapidly emerging
Zuzana Machacova et al.
Nature communications, 15(1), 7375-7375 (2024-08-28)
PARP inhibitors (PARPi), known for their ability to induce replication gaps and accelerate replication forks, have become potent agents in anticancer therapy. However, the molecular mechanism underlying PARPi-induced fork acceleration has remained elusive. Here, we show that the first PARPi-induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service