Skip to Content
Merck
All Photos(1)

Documents

C4464

Sigma-Aldrich

EZMix N-Z-Amine® A

Casein enzymatic hydrolysate, from bovine milk

Synonym(s):

Peptone from casein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352116
NACRES:
NA.85

biological source

bovine milk

grade

for molecular biology

sterility

non-sterile

form

powder

application(s)

agriculture
(ffff123)

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

Looking for similar products? Visit Product Comparison Guide

General description

NZ-Amine® EKC is derived from the enzymatic digest of casein, often used in culture media. NZ Amine EKC contains a mixture of peptides and amino acids.

Application

EZMix N-Z-Amine® A has been used as a nutritional source in microbial media and plant cell suspension cultures.
EZMix N-Z-Amine® A is suitable for use as a nutritional source in microbial media.

Features and Benefits

NZ Amine EZMix powder provides:
  • Granulated, dust-free format for safer handling and faster mixing
  • Easy scale-up using larger package sizes

Legal Information

N-Z-Amine is a registered trademark of Kerry Group

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jens C Benninghoff et al.
Applied and environmental microbiology, 87(11) (2021-03-21)
Sulfolobus acidocaldarius is a thermoacidophilic crenarchaeon with optimal growth at 80°C and pH 2 to 3. Due to its unique physiological properties, allowing life at environmental extremes, and the recent availability of genetic tools, this extremophile has received increasing interest
Michael L Reno et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(21), 8605-8610 (2009-05-14)
Variation in gene content has been hypothesized to be the primary mode of adaptive evolution in microorganisms; however, very little is known about the spatial and temporal distribution of variable genes. Through population-scale comparative genomics of 7 Sulfolobus islandicus genomes
Natali R Mustafa et al.
Nature protocols, 6(6), 715-742 (2011-06-04)
Methods described in this paper are confined to in vitro dedifferentiated plant cell suspension cultures, which are convenient for the large-scale production of fine chemicals in bioreactors and for the study of cellular and molecular processes, as they offer the
F R Blattner et al.
Science (New York, N.Y.), 196(4286), 161-169 (1977-04-08)
The Charon lambda bacteriophages have been developed as vectors for cloning. Their construction incorporates mutations that make them simple to use and also greatly increases their safety for the biological containment of cloned recombinant DNA. Three of the Charon vector
Adam B Robertson et al.
Nature protocols, 7(2), 340-350 (2012-01-28)
We describe a method for the efficient and selective identification of DNA containing the 5-hydroxymethylcytosine (5-hmC) modification. This protocol takes advantage of two proteins: T4 β-glucosyltransferase (β-gt), which converts 5-hmC to β-glucosyl-5-hmC (β-glu-5-hmC), and J-binding protein 1 (JBP1), which specifically

Protocols

General protocols for growth of competent cells in microbial medium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service