Skip to Content
Merck
All Photos(1)

Key Documents

C4396

Sigma-Aldrich

Carbonic Anhydrase I from human erythrocytes

Synonym(s):

Carbonate Dehydratase, Carbonate Hydrolyase, Carbonic Anhydrase Isozyme I

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.25

form

powder

specific activity

100-500 W-A units/mg protein

mol wt

30 kDa

color

white

pI 

~6.6

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CA1(759)

Looking for similar products? Visit Product Comparison Guide

Application

Carbonic anhydrase from human erythrocytes (HCA) has been used to study the molten-globule state of carbonic anhydrase (CA). Chaperone-like α-crystallin binds to this state of the enzyme and prevents its aggregation. The enzyme from sigma has been used for the analysis of thermodynamic stability of the enzyme. Furthermore, its clinical significance has been evaluated in human non-small cell lung cancer.

Biochem/physiol Actions

Carbonic anhydrase is a zinc metalloenzyme that has a molecular weight of approximately 30,000 Da. The enzyme catalyzes the hydration of carbon dioxide to carbonic acid. It is involved in vital processes such as pH and CO2 homeostasis, transport of bicarbonate and CO2, biosynthetic reactions, bone resorption, calcification, and tumorigenicity. Therefore, this enzyme is an important target for inhibitors with clinical applications in various pathologies such as glaucoma, epilepsy and Parkinson′s disease.

Unit Definition

One Wilbur-Anderson (W-A) unit will cause the pH of a 0.02 M Trizma buffer to drop from 8.3 to 6.3 per min at 0 °C. (One W-A unit is essentially equivalent to one Roughton-Booth unit.)

Preparation Note

chromatographically
purified.

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Rajaraman et al.
The Journal of biological chemistry, 271(44), 27595-27600 (1996-11-01)
alpha-Crystallin, a multimeric protein, exhibits chaperone-like activity in preventing aggregation of several proteins. We have studied the chaperone-like activity of alpha-crystallin toward heat-induced aggregation of bovine and human carbonic anhydrase. Human carbonic anhydrase aggregates at 60 degrees C, while bovine
Whei Ling Chiang et al.
Cancer letters, 188(1-2), 199-205 (2002-10-31)
This study was designed to elucidate the possible relationship between the expression of cytosolic carbonic anhydrase (CA) and non-small cell lung cancer (NSCLC). The activity and protein expression patterns of carbonic anhydrase I (CAI) and II (CAII) of 70 NSCLC
Pavel V Shliaha et al.
Journal of proteome research, 12(6), 2323-2339 (2013-03-22)
qTOF mass spectrometry and traveling wave ion mobility separation (TWIMS) hybrid instruments (q-TWIMS-TOF) have recently become commercially available. Ion mobility separation allows an additional dimension of precursor separation inside the instrument, without incurring an increase in instrument time. We comprehensively
Daumantas Matulis et al.
Biochemistry, 44(13), 5258-5266 (2005-03-30)
ThermoFluor (a miniaturized high-throughput protein stability assay) was used to analyze the linkage between protein thermal stability and ligand binding. Equilibrium binding ligands increase protein thermal stability by an amount proportional to the concentration and affinity of the ligand. Binding
Qiang Jin et al.
Biosensors & bioelectronics, 83, 193-199 (2016-04-30)
A near-infrared fluorescent probe (DDAB) for highly selective and sensitive detection of carboxylesterase 2 (CE2) has been designed, synthesized, and systematically studied both in vitro and in vivo. Upon addition of CE2, the ester bond of DDAB could be rapidly

Protocols

Objective: To standardize a procedure for the enzymatic assay of Carbonic Anhydrase (EC 4.2.1.1) for Wilbur-Anderson Units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service