Skip to Content
Merck

C4142

Capreomycin sulfate from Streptomyces capreolus

antibacterial peptide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.85
PubChem Substance ID:
UNSPSC Code:
51281628
EC Number:
215-776-8
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria, mycobacteria

mode of action

DNA synthesis | interferes

storage temp.

−20°C

SMILES string

OS(O)(=O)=O.NCCC[C@H](N)C(=O)NC[C@H]1NC(=O)[C@H](CO)NC(=O)[C@H](N)CNC(=O)[C@@H](NC(=O)\C(NC1=O)=C\NC(N)=O)[C@H]2CCNC(=N)N2

InChI

1S/C24H42N14O8.H2O4S/c25-4-1-2-10(26)17(40)32-7-13-19(42)34-14(8-33-24(29)46)20(43)38-16(12-3-5-30-23(28)37-12)22(45)31-6-11(27)18(41)36-15(9-39)21(44)35-13;1-5(2,3)4/h8,10-13,15-16,39H,1-7,9,25-27H2,(H,31,45)(H,32,40)(H,34,42)(H,35,44)(H,36,41)(H,38,43)(H3,28,30,37)(H3,29,33,46);(H2,1,2,3,4)/b14-8-;/t10-,11+,12?,13+,15-,16-;/m0./s1

InChI key

LFFNIXQXRKNZCE-UBBQZPMLSA-N

Application

Capreomycin is used to study bacterial ribosomal subunit interactions and translocation processes during protein synthesis. It is a second line antibiotic and is used to study multidrug-resistant tuberculosis.

Biochem/physiol Actions

Capreomycin is a cyclic peptide antibiotic that is often grouped with aminoglycosides. It binds across the ribosomal interface involving 23S rRNA helix 69 (H69) and 16S rRNA helix 44 (h44).

Preparation Note

Sparingly soluble in water.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library