Skip to Content
Merck
All Photos(3)

Key Documents

C4011

Sigma-Aldrich

Cytochrome c from pigeon breast muscle

≥95% based on Mol. Wt. 12,173 basis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

pigeon muscle (breast)

Quality Level

Assay

≥95% based on Mol. Wt. 12,173 basis

form

powder

technique(s)

cell culture | mammalian: suitable

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Cytochrome c has been identified as an important mediator in apoptotic pathways. The release of mitochondrial cytochrome c into the cytoplasm stimulates apoptosis and is commonly used as an indicator of the apoptotic process in the cell. Investigation on the effect of Paris Saponin I (PS I) on human gastric carcinoma cell growth (SGC7901 cells) have shown an elevated level cytoplasmic cytochrome c. Results are an inhibition of proliferation in SGC7901 cells by inducing mitochondria-dependent apoptosis through cytochrome c.

Biochem/physiol Actions

The ready fluctuation of cytochrome c within the cell between ferrous and ferric states, makes it an efficient biological electron-transporter. It plays a vital role in cellular oxidations in both plants and animals. Generally regarded as a universal catalyst of respiration, it forms the essential electron-bridge between the respirable substrates and oxygen.

Preparation Note

Prepared with acetic acid without using TCA.

Other Notes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin-Lin Gao et al.
World journal of gastroenterology, 17(39), 4389-4395 (2011-11-24)
To investigate the anti-tumor effects of Paris chinensis dioscin (PCD) and mechanisms regarding cell cycle regulation and apoptosis in human gastric cancer SGC-7901 cells. Cell viability was analyzed by the 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl-tetrazolium bromide assay. Cell apoptosis was evaluated by flow cytometry
Esther Lapuente-Brun et al.
Science (New York, N.Y.), 340(6140), 1567-1570 (2013-07-03)
The textbook description of mitochondrial respiratory complexes (RCs) views them as free-moving entities linked by the mobile carriers coenzyme Q (CoQ) and cytochrome c (cyt c). This model (known as the fluid model) is challenged by the proposal that all
Xinliu Gao et al.
Archives of biochemistry and biophysics, 535(2), 197-204 (2013-04-17)
The filamentous anoxygenic phototrophic bacterium Chloroflexus aurantiacus possesses an unusual electron transfer complex called Alternative Complex III instead of the cytochrome bc or bf type complex found in nearly all other known groups of phototrophs. Earlier work has confirmed that
Rawabi Qadhi et al.
Toxicology letters, 219(3), 288-297 (2013-03-26)
The importance of dietary polyunsaturated fatty acids (PUFAs) in the reduction of cardiovascular disease has been recognized for many years. Docosahexaenoic acid (22:6n3, DHA) is an n-3 PUFA known to affect numerous biological functions and provide cardioprotection; however, the exact
Eva Di Silvio et al.
Biochimica et biophysica acta, 1834(8), 1554-1561 (2013-05-08)
The biogenesis of c-type cytochromes (Cytc) is a process that in Gram-negative bacteria demands the coordinated action of different periplasmic proteins (CcmA-I), whose specific roles are still being investigated. Activities of Ccm proteins span from the chaperoning of heme b

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service