Skip to Content
Merck
All Photos(2)

Key Documents

C2970

Sigma-Aldrich

Monoclonal Anti-Cathepsin L antibody produced in mouse

clone 33/2, ascites fluid

Synonym(s):

Anti-CATL, Anti-CTSL, Anti-MEP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

33/2, monoclonal

mol wt

antigen (human cathepsin L) 25 kDa
antigen (human procathepsin L) 42 kDa

contains

15 mM sodium azide

species reactivity

mouse, mink, human, rat

technique(s)

immunohistochemistry (frozen sections): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 1:200 using whole cell extract of a cultured mink lung cell line

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTSL1(1514)
mouse ... Ctsl(13039)
rat ... Ctsl1(25697)

Looking for similar products? Visit Product Comparison Guide

General description

Cathepsin L is a cysteine protease and is secreted by numerous transformed cells in its inactive pre-form. It is ubiquitously expressed and is localized in nucleus. Cathepsin L have substrate interacting region located in L and R domain loops.

Specificity

Reacts specifically with native and denatured forms of human cathepsin L and procathepsin L. Recognizes an epitope within amino acid residues GYGFEST (265-271 in procathepsin L and 169-175 in the mature cathepsin L molecule). The antibody may be used for the suppression of the malignant growth of myeloma cells. Cross-reactivity is observed with rat cathepsin L (strong), mouse procathepsin L (weak) and mink cathepsin and procathepsin L. Does not cross-react with human cathepsin types B, D, H and S, procathepsins B and D, rat cathepsin B, and mouse procathepsins B and D.

Immunogen

procathepsin L isolated from the human lung cancer cell line EPLC 32M1.

Biochem/physiol Actions

Cathepsin L expression is correlated with the metastatic potential of transformed cells. Cathepsin L is capable of degrading protein constituents of the extracellular matrix, this enzyme is thought to play a crucial role in tumor progression, metastasis and other disorders where the destruction of the matrix is the major cause of disease.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cysteine cathepsins: from structure, function and regulation to new frontiers
Turk V, et al
Biochimica et Biophysica Acta (BBA)-Proteins and Proteomics, 1824(1), 68-88 (2012)
Cathepsin-L influences the expression of extracellular matrix in lymphoid organs and plays a role in the regulation of thymic output and of peripheral T cell number
Lombardi G, et al.
Journal of Immunology, 174(11), 7022-7032 (2005)
Kazuyoshi Yanagihara et al.
Cancer science, 96(6), 323-332 (2005-06-17)
The number of published studies on peritoneal dissemination of scirrhous gastric carcinoma is very small as a result of the unavailability of highly reproducible animal models. Orthotopic implantation of HSC-44PE and HSC-58 (scirrhous gastric carcinoma-derived cell lines) cells into nude
Increased expression and activity of nuclear cathepsin L in cancer cells suggests a novel mechanism of cell transformation
Goulet B, et al.
Molecular Cancer Research, 5(9), 899-907 (2007)
Cathepsin L1, the Major Protease Involved in Liver Fluke (Fasciola hepatica) virulence propeptide cleavage sites and autoactivation of the zymogen secreted from gastrodermal cells
Collins PR, et al.
Test, 279(17), 17038-17046 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service