C-28019
Mesenchymal Stem Cell Growth Medium DXF
Ready-to-use kit including Basal Medium and SupplementMix, 500 ml
About This Item
Recommended Products
form
liquid
packaging
pkg of 500 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
General description
Application
Quality
Storage and Stability
Subculture Routine
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Protocol to differentiate human mesenchymal stem cells into neurons with extensive dendrites and axons in 3 days. Explore over 350 PromoCell products on Sigma.com.
Protocol to differentiate human mesenchymal stem cells into neurons with extensive dendrites and axons in 3 days. Explore over 350 PromoCell products on Sigma.com.
Are your mesenchymal stem cells really MSCs? Learn more about the MSC characterization guidelines proposed by the International Society for Cellular Therapy (ISCT).
Are your mesenchymal stem cells really MSCs? Learn more about the MSC characterization guidelines proposed by the International Society for Cellular Therapy (ISCT).
Protocols
Mesenchymal Stem Cells and Media enable in vitro expansion and differentiation, vital for various tissue sources.
Mesenchymal Stem Cells and Media enable in vitro expansion and differentiation, vital for various tissue sources.
Mesenchymal Stem Cells and Media enable in vitro expansion and differentiation, vital for various tissue sources.
Mesenchymal Stem Cells and Media enable in vitro expansion and differentiation, vital for various tissue sources.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service