Skip to Content
Merck
All Photos(1)

Key Documents

C0993

Sigma-Aldrich

CGP 57380

≥98% (HPLC)

Synonym(s):

CGP57380, N3-(4-fluorophenyl)-1h-pyrazolo[3,4-d]pyrimidine-3,4-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9FN6
CAS Number:
Molecular Weight:
244.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear
H2O: <2 mg/mL

storage temp.

2-8°C

SMILES string

Nc1ncnc2[nH]nc(Nc3ccc(F)cc3)c12

InChI

1S/C11H9FN6/c12-6-1-3-7(4-2-6)16-11-8-9(13)14-5-15-10(8)17-18-11/h1-5H,(H4,13,14,15,16,17,18)

InChI key

UQPMANVRZYYQMD-UHFFFAOYSA-N

Application

CGP 57380 has been used:
  • as a mitogen-activated protein kinase-interacting kinase 1 (MNK1) inhibitor to investigate the role of Mnk1 on eukaryotic translation initiation factor 4F (eIF4F) assembly and Newcastle disease virus (NDV) replication in HeLa cells
  • as an MNK1 inhibitor to block eIF4 complex to evaluate the contribution of interferon (IFN-γ) translation during m157- transgenic (Tg) stimulation
  • as a β-catenin nuclear translocation inhibitor to analyze its effects on cytoplasmic and nuclear fractions of cells

Biochem/physiol Actions

CGP 57380 is a β-catenin nuclear translocation inhibitor, which prevents the proliferation of several cancers. It enhances radiation-induced apoptosis in nasopharyngeal carcinoma (NPC). CGP 57380 upregulates β-catenin in the cytoplasm and inhibits epithelial-mesenchymal transition (EMT).
CGP 57380 is a cell-permeable selective inhibitor of mitogen-activated protein kinase-interacting kinase 1 (MNK1).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W-C Huangfu et al.
Oncogene, 31(2), 161-172 (2011-06-15)
Interferon alpha (IFNα) is widely used for treatment of melanoma and certain other malignancies. This cytokine as well as the related IFNβ exerts potent anti-tumorigenic effects; however, their efficacy in patients is often suboptimal. Here, we report that inflammatory signaling
David Müller et al.
Translation (Austin, Tex.), 1(2), e25819-e25819 (2013-01-01)
In eukaryotes, mRNA translation is dependent on the cap-binding protein eIF4E. Through its simultaneous interaction with the mRNA cap structure and with the ribosome-associated eIF4G adaptor protein, eIF4E physically posits the ribosome at the 5' extremity of capped mRNA. eIF4E
Michael Schümann et al.
Cancer research, 66(3), 1282-1288 (2006-02-03)
The Raf/mitogen-activated protein/extracellular signal-regulated kinase (ERK) kinase (MEK)/ERK signaling cascade enhances tumor cell proliferation in many cases. Here, we show that adenovirus type 5, a small DNA tumor virus used in experimental cancer therapy, strongly induces ERK phosphorylation during the
Yeon-Ho Chung et al.
Scientific reports, 6, 34533-34533 (2016-10-04)
IL-1β is a key mediator of sterile inflammation in response to endogenous particulates, a type of damage-associated molecular pattern (DAMPs) molecule derived from damaged cells. Despite the well-known role of sterile particulates such as monosodium urate (MSU) crystals as inflammasome
L S D'Abronzo et al.
Oncogene, 36(46), 6359-6373 (2017-07-27)
The antiandrogen bicalutamide is widely used in the treatment of advanced prostate cancer (PCa) in many countries, but its effect on castration-resistant PCa (CRPC) is limited. We previously showed that resistance to bicalutamide results from activation of mechanistic target of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service