Skip to Content
Merck

Skip To

C0712

CDP-Star® Chemiluminescent Substrate

0.25 mM in H2O

Synonym(s):

Disodium 2-chloro-5-(4-methoxyspiro[1,2-dioxetane-3,2′-(5-chlorotricyclo[3.3.1.13.7]decan])-4-yl]-1-phenyl phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H19Cl2O7Na2P
CAS Number:
Molecular Weight:
495.20
NACRES:
NA.83
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥75% (HPLC)

Quality Segment

form

liquid

usage

sufficient for 20 blots detection of alkaline phosphatase-labeled molecules (10 × 10 cm)

concentration

0.25 mM in H2O

storage temp.

2-8°C

SMILES string

Cl[C@]1(C2)C[C@H]3C4(OOC4(C5=CC(OP([O-])([O-])=O)=C(Cl)C=C5)OC)[C@@H]2C[C@@H](C1)C3

InChI

1S/C18H21Cl2O7P.2Na/c1-24-18(14-6-13(2-3-15(14)19)25-28(21,22)23)17(26-27-18)11-4-10-5-12(17)9-16(20,7-10)8-11;;/h2-3,6,10-12H,4-5,7-9H2,1H3,(H2,21,22,23);;/q;2*+1/p-2/t10-,11-,12+,16-,17?,18?;;

InChI key

UKWLRLAKGMZXJC-VCCHNUGISA-L

Application

CDP-Star® is a sensitive, chemiluminescent substrate for alkaline phosphatase that allows for the rapid, reproducible detection of alkaline phosphatase-labeled molecules in Northern, Southern, and Western blotting applications. Detection of alkaline phosphatase-labeled molecules with CDP-Star is extremely sensitive as a result of low background luminescence coupled with high intensity and prolonged light output from the enzyme catalysis. Following exposure to alkaline phosphatase-labeled molecules, maximum light emission occurs at approximately 60 minutes and continues for up to 24 hours, allowing for multiple film exposures and/or the sensitive detection of targets present in small amounts. For convenience, CDP-Star is supplied as a 0.25 mM, ready-to-use aqueous solution (i.e. no dilution is necessary). CDP-Star functions on both neutral and positively-charged nylon, giving the reagent added application versatility.

Features and Benefits

CDP-Star® is supplied as a 0.25 mM ready-to-use aqueous solution for use in a variety of membrane-based applications. CDP-Star functions on both neutrally-charged and positively-charged nylon, giving the reagent added application versatility.

Legal Information

CDP-Star® is a trademark of Tropix, Inc. Bedford, MA, USA and covered under U.S. Patent 5,326,882.
CDP-Star is a registered trademark of Tropix, Inc.

Disclaimer

Store at 2-8 °C protected from light.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CDP-ROSML3379N2790
assay

≥75% (HPLC)

assay

>98%

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

200

Quality Level

-

Quality Level

100

Quality Level

100

form

liquid

form

solution

form

powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

usage

sufficient for 20 blots detection of alkaline phosphatase-labeled molecules (10 × 10 cm)

usage

-

usage

-

usage

-

concentration

0.25 mM in H2O

concentration

-

concentration

-

concentration

-


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters