Skip to Content
Merck

C0330

Ciprofibrate

Synonym(s):

2-[p-(2,2-Dichlorocyclopropyl)phenoxy]-2-methylpropanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H14Cl2O3
CAS Number:
Molecular Weight:
289.15
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
257-744-6
MDL number:
eCl@ss:
32150313
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

storage temp.

2-8°C

SMILES string

CC(C)(Oc1ccc(cc1)C2CC2(Cl)Cl)C(O)=O

InChI

1S/C13H14Cl2O3/c1-12(2,11(16)17)18-9-5-3-8(4-6-9)10-7-13(10,14)15/h3-6,10H,7H2,1-2H3,(H,16,17)

InChI key

KPSRODZRAIWAKH-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Biochem/physiol Actions

Peroxisome proliferator-activated receptor α (PPARα) agonist


Still not finding the right product?

Explore all of our products under Ciprofibrate


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Precautionary Statements

Hazard Classifications

Carc. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library