Skip to Content
Merck
All Photos(1)

Key Documents

C0128

Sigma-Aldrich

α-Chloralose

Synonym(s):

1,2-O-(2,2,2-Trichloroethylidene)-α-D-glucofuranose, Anhydro-D-glucochloral, Chloralose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11Cl3O6
CAS Number:
Molecular Weight:
309.53
Beilstein:
85418
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

form

powder

Quality Level

impurities

≤20% β-anomer

color

white to off-white

solubility

ethanol: 10 mg/mL, clear to slightly hazy, colorless to faintly yellow (or yellow-brown)

SMILES string

OC[C@@H](O)[C@H]1O[C@@H]2O[C@@H](O[C@@H]2[C@H]1O)C(Cl)(Cl)Cl

InChI

1S/C8H11Cl3O6/c9-8(10,11)7-16-5-3(14)4(2(13)1-12)15-6(5)17-7/h2-7,12-14H,1H2/t2-,3+,4-,5-,6-,7-/m1/s1

InChI key

OJYGBLRPYBAHRT-IPQSZEQASA-N

Looking for similar products? Visit Product Comparison Guide

Application

α-Chloralose has been used as anesthetic in surgical preparations.

Biochem/physiol Actions

α-Chloralose is used as an immobilizing agent. It is mostly used in studies, which involves autonomic reflexes. α-Chloralose is a sedative hypnotic. It has high significance as anesthetic for laboratory animals.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serotonin in the solitary tract nucleus shortens the laryngeal chemoreflex in anaesthetized neonatal rats
Donnelly WT, et al.
Experimental Physiology, 101(7), 946-961 (2016)
Preparation of water-soluble compounds by covalent attachment of solubilizing moieties
The Practice of Medicinal Chemistry, 767-785 (2008)
Activation of serotonergic neurons in the medullary caudal raphe shortens the laryngeal chemoreflex in anaesthetized neonatal rats
Donnelly WT, et al.
Experimental Physiology, 102(8), 1007-1018 (2017)
Vertebrate pest control chemicals and their use in urban and rural environments
Hayes' Handbook of Pesticide Toxicology, 271-284 (2010)
Preanesthesia, anesthesia, analgesia, and euthanasia
Laboratory Animal Medicine, 1135-1200 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service