Skip to Content
Merck
All Photos(4)

Key Documents

C0125

Sigma-Aldrich

Cacodylic acid

≥98%

Synonym(s):

Dimethylarsinic acid, Dimethylarsonic acid, Hydroxydimethylarsine oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2As(O)OH
CAS Number:
Molecular Weight:
138.00
Beilstein:
1736965
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98%

form

powder

solubility

water: 50 mg/mL, clear, colorless

SMILES string

C[As](C)(O)=O

InChI

1S/C2H7AsO2/c1-3(2,4)5/h1-2H3,(H,4,5)

InChI key

OGGXGZAMXPVRFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monika K Nisiewicz et al.
International journal of molecular sciences, 22(1) (2021-01-08)
Nearly half of patients with advanced and metastatic melanomas harbor a BRAF mutation. Vemurafenib (VEM), a BRAF inhibitor, is used to treat such patients, however, responses to VEM are very short-lived due to intrinsic, adaptive and/or acquired resistance. In this
Fang-Chih Chang et al.
Talanta, 198, 137-145 (2019-03-17)
An analytical method for the biomonitoring of arsenic, benzene and polycyclic aromatic hydrocarbons (PAHs) in human urine was developed using liquid chromatography tandem mass spectrometry (LC-MS/MS). The urinary metabolites of monomethylarsonic acid (MMAA), dimethylarsonic acid (DMAA), s-phenylmercapturic acid (S-PMA), and
Erik J Tokar et al.
Toxicology letters, 209(2), 179-185 (2012-01-11)
Inorganic arsenic, an early life carcinogen in humans and mice, can initiate lesions promotable by other agents in later life. The biomethylation product of arsenic, dimethylarsinic acid (DMA), is a multi-site tumor promoter. Thus, pregnant CD1 mice were given drinking
Hua Naranmandura et al.
Toxicological sciences : an official journal of the Society of Toxicology, 128(1), 137-146 (2012-04-28)
Arsenic is known to be a human carcinogen as well as one of the most effective drugs for treatment of patients with acute promyelocytic leukemia. The intermediate metabolites of arsenic, monomethylarsonous acid (MMA(III)) and dimethylarsinous acid (DMA(III)), are formed by
S Yamamoto et al.
Mutation research, 386(3), 353-361 (1997-06-01)
The modifying effects of dimethylarsinic acid (DMA), the major metabolite of ingested arsenicals in most mammals, on chemical carcinogenesis were investigated using rat in vivo models and reviewed here. In a multi-organ bioassay, rats pretreated with 5 carcinogens were administered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service