BR780536
BRAND® microcentrifuge tube, 0.5 mL with lid
clear polypropylene
All Photos(1)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
Recommended Products
material
clear polypropylene
sterility
non-sterile
manufacturer/tradename
BRAND
O.D. × H
10 mm × 30 mm
Related Categories
General description
The disposable microtubes with 0.5 ml nominal volume are made from high-quality PP. The quality level BIO-CERT® PCR QUALITY ensures the required cleanliness for PCR-setups, storage of PCR samples and other applications with high cleanliness requirements.They are available with an easy-to-open snap-on lid in versions with or without lid locking. Lid locking prevents unintended opening of the lid when heating the samples inside the microtubes. The disposable microtubes can be centrifuged up to the maximum RCF value stated and autoclaved at 121 °C (2 bar) according to DIN EN 285. For a reading of the approximate volume, they have an easy-to-read graduation.BIO-CERT® PCR QUALITY: Free of human DNA, RNase, DNase, Pyrogens and PCR-Inhibitors
Features and Benefits
- High cleanliness for demanding applications
- Tight sealing snap lid
- Can be opened and closed with one hand
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)