Skip to Content
Merck
All Photos(1)

Key Documents

B9688

Sigma-Aldrich

Pizotifen

≥98% (HPLC)

Synonym(s):

Pizotifen, Pizotyline; 4-(9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NS
CAS Number:
Molecular Weight:
295.44
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: ≥8 mg/mL

originator

Novartis

storage temp.

room temp

SMILES string

CN1CCC(\CC1)=C2\c3ccccc3CCc4sccc24

InChI

1S/C19H21NS/c1-20-11-8-15(9-12-20)19-16-5-3-2-4-14(16)6-7-18-17(19)10-13-21-18/h2-5,10,13H,6-9,11-12H2,1H3

InChI key

FIADGNVRKBPQEU-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

Pizotifen is a Serotonin antagonist used for prophylaxis of migraine
Pizotifen is a serotonin antagonist acting mainly at the 5-HT1, 5-HT2A and 5HT2C receptors with some antihistamine activity. It is used for the prevention of vascular headache including migraine and cluster headache.

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C E Serna-Jiménez et al.
Biomedical chromatography : BMC, 26(6), 769-774 (2011-10-18)
Pizotifen malate is an antihistamine and serotonin inhibitor used in the preventive treatment of migraine and eating disorders. A simple, rapid, accurate and precise high-performance liquid chromatography (HPLC) method involving ultraviolet detection was validated for the quantitative analysis of pizotifen
Richard Young et al.
Pharmacology, biochemistry, and behavior, 82(2), 404-410 (2005-10-29)
MDMA (N-methyl-3,4-methylenedioxyamphetamine) produces a discriminative stimulus (DS) effect in animals, but attempts to completely block this action with selective neurotransmitter antagonists have not been very successful. Biochemically, MDMA can increase synaptic levels of serotonin, dopamine, and norepinephrine that, conceivably, might
M H Guillet et al.
Annales de dermatologie et de venereologie, 122(11-12), 777-779 (1995-01-01)
Primary erythermalgia is a rare acrosyndrome due to paroxystic arteriolar dilatation of extremities that become red and congested with warmth and burning sensation or pain. We report herein the observation of a primary erythermalgia affecting 3 degrees in the same
Paul Silcocks et al.
Trials, 11, 71-71 (2010-06-18)
A recent Cochrane Review demonstrated the remarkable lack of reliable clinical trials of migraine treatments for children, especially for the two most prescribed preventative treatments in the UK, Propranolol and Pizotifen.Migraine trials in both children and adults have high placebo
Abdeslam Chagraoui et al.
Psychopharmacology, 170(1), 17-22 (2003-08-09)
Agomelatine, an antidepressant with melatonin agonist and 5-HT(2C) antagonist properties, as well as two of its main metabolites, S 21517 (N-[2-(7-hydroxy-1-naphtyl)ethyl]acetamide) and S21540 (N-[2-(3-hydroxy-7-methoxynaphtalen-1-yl)ethyl]acetamide), have been assessed in vitro on pig choroid plexus preparations to determine their affinities for 5-HT(2C)

Articles

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service