Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
form
tablet
Quality Level
solubility
deionized water: one tablet, 5 mg/mL
storage temp.
−20°C
SMILES string
[Na+].O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C([O-])=O)Oc2c[nH]c3ccc(Br)c(Cl)c23
InChI
1S/C14H13BrClNO7.Na/c15-4-1-2-5-7(8(4)16)6(3-17-5)23-14-11(20)9(18)10(19)12(24-14)13(21)22;/h1-3,9-12,14,17-20H,(H,21,22);/q;+1/p-1/t9-,10-,11+,12-,14+;/m0./s1
InChI key
IBLSVGDGSKUDCT-CYRSAHDMSA-M
Application
Preparation Note
1 of 1
This Item | |||
|---|---|---|---|
| solubility deionized water: one tablet, 5 mg/mL | solubility - | solubility water: 50 mg/mL, clear, colorless | solubility - |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| form tablet | form tablet | form powder | form tablet |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Still not finding the right product?
Explore all of our products under 5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| B8174-5TAB | 04061826588963 |


