Skip to Content
Merck
All Photos(3)

Documents

B4527

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl β-D-glucopyranoside

chromogenic, ≥97%, powder

Synonym(s):

X-Glc, X-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
Beilstein:
1552603
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

5-Bromo-4-chloro-3-indolyl β-D-glucopyranoside, ≥97%

Assay

≥97%

form

powder

solubility

DMF: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2c[nH]c3ccc(Br)c(Cl)c23)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C14H15BrClNO6/c15-5-1-2-6-9(10(5)16)7(3-17-6)22-14-13(21)12(20)11(19)8(4-18)23-14/h1-3,8,11-14,17-21H,4H2/t8-,11-,12+,13-,14-/m1/s1

InChI key

OPIFSICVWOWJMJ-LNNRFACYSA-N

Looking for similar products? Visit Product Comparison Guide

Substrates

A histochemical substrate for β-glucosidase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hien P Nguyen et al.
Genes, 11(5) (2020-05-01)
Bradyrhizobium elkanii USDA61 possesses a functional type III secretion system (T3SS) that controls host-specific symbioses with legumes. Here, we demonstrated that B. elkanii T3SS is essential for the nodulation of several southern Asiatic Vigna mungo cultivars. Strikingly, inactivation of either
L A Shelef et al.
Letters in applied microbiology, 25(3), 202-206 (1997-11-14)
A new instrument, capable of detecting metabolic changes due to microbiological activity, is described. Optical changes in growth media are monitored in a semi-fluid zone that separates the liquid medium containing the sample. Data demonstrate that common media can be
W D Watkins et al.
Applied and environmental microbiology, 54(7), 1874-1875 (1988-07-01)
A new chromogenic compound, 5-bromo-4-chloro-3-indoxyl-beta-D-glucuronide, was found to be useful for the rapid, specific, differential identification of Escherichia coli in the sanitary analysis of shellfish and wastewater. Of 1,025 presumptively positive colonies (blue) and 583 presumptively negative colonies (nonblue), only
H Arakawa et al.
Analytical biochemistry, 199(2), 238-242 (1991-12-01)
Chemiluminescent assays of various enzymes have been developed using indoxyl derivatives as substrates. The principle of the method is as follows: an enzyme causes hydrolysis of an indoxyl derivative to an intermediate indoxyl that is readily oxidized to indigo dye
R Gossrau et al.
Histochemistry, 86(4), 397-404 (1987-01-01)
5-Br-4-Cl-3-Indoxyl-alpha-D-gluco(pyrano)side was found to be the most suitable synthetic substrate for the demonstration of alpha-D-glucosidases in situ. Using an azoindoxyl procedure with hexazotized pararosaniline or new fuchsine at pH 5 in freeze-dried celloidine-mounted cryostat sections acid alpha-D-glucosidase (EC 3.2.1.20) was

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service