Skip to Content
Merck
All Photos(1)

Key Documents

B3648

Sigma-Aldrich

Anti-Human IgG4−Biotin antibody, Mouse monoclonal

clone HP-6025, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Human IgG4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

HP-6025, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

direct ELISA: 1:60,000

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

Specificity

Monoclonal Anti-Human IgG4 is specific for the IgG4 subclass and nonreactive with IgG1, IgG2 and IgG3 in an ELISA.

Application

Anti-Human IgG4−Biotin antibody, Mouse monoclonal has been used in immunohistochemistry and immunostaining.
Monoclonal Anti-Human IgG4-Biotin antibody produced in mouse may be used in ELISA at a working dilution of 1:15,000. It was used for detection of serum IgG4 produced in response to malaria parasite infection by ELISA.
The IUIS/WHO2 study singled out this monoclonal antibody as one of the most widely applicable IgG4 specific monoclonal antibodies. Monoclonal Anti-Human IgG4 may be used for the identification of the IgG4 subclass by various immunoassays including: ELISA, Imprint Immunofixation (IIF), Immunofluorometric Assay (IFMA), Hemagglutination (HA), Hemagglutination Inhibition (HAI), Particle Counting Immunoassay (PACIA), and detection of cytoplasmic IgG.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of biotin to monoclonal Anti-Human IgG4 antibody allows for the binding of various labels such as avidin or streptavidin.
IgG4 is the least abundant immunoglobulin present in the human serum. IgG4 is implicated in a unique process called fab-arm exchange (FAE) and limits the formation of immune complex. It has anti-inflammatory properties. IgG4 functions as a therapeutic antibody. It provides protection against allergy, prevents mast cell degranulation and has a harmful role in malignant melanoma. IgG4 may be associated with autoimmune diseases such as, autoimmune pancreatitis (AIP) type 1 and 2, Mikulicz syndrome, Kuttner′s tumor, Riedel′s thyroiditis and tubulointerstitial nephritis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hayder A Giha et al.
Malaria journal, 8, 306-306 (2009-12-24)
Plasmodium falciparum malaria is a complex disease in which genetic and environmental factors influence susceptibility. IgG isotypes are in part genetically controlled, and GM/KM allotypes are believed to be involved in this control. In this study, 216 individuals from Daraweesh
Intranasal air sampling in homes: relationships among reservoir allergen concentrations and asthma severity
Gore RB, et al.
The Journal of Allergy and Clinical Immunology, 117(3), 649-655 (2006)
A Nasr et al.
Scandinavian journal of immunology, 74(4), 390-396 (2011-06-08)
The anti-malarial IgG immune response during the lengthy and dry season in areas of low malaria transmission as in Eastern Sudan is largely unknown. In this study, ELISA was used for the measurement of pre-existing total IgG and IgG subclasses
A Nasr et al.
Scandinavian journal of immunology, 66(1), 87-96 (2007-06-26)
In a prospective clinical study in New Halfa Teaching Hospital, the possible association between FcgammaRIIa-R/H131 polymorphism and anti-malarial antibody responses with clinical outcome of Plasmodium falciparum malaria among Sudanese patients was investigated. A total of 256 individuals were consecutively enrolled
IgG4-related autoimmune diseases: Polymorphous presentation complicates diagnosis and treatment
Kleger A, et al.
Deutsches Arzteblatt International, 112(8), 128-128 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service