Skip to Content
Merck
All Photos(1)

Key Documents

B0261

Sigma-Aldrich

Bufalin

Synonym(s):

3β,14-Dihydroxy-5β,20(22)-bufadienolide, 5β,20(22)-Bufadienolide-3β,14-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34O4
CAS Number:
Molecular Weight:
386.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥97% (HPLC)

Quality Level

form

solid

solubility

DMF: soluble
DMSO: soluble
ethanol: soluble

storage temp.

2-8°C

SMILES string

O[C@]([C@@](CC[C@@]1([H])[C@@]2(CC[C@H](O)C1)C)([H])[C@]2([H])CC3)(CC[C@@H]4C(C=C5)=COC5=O)[C@]34C

InChI

1S/C24H34O4/c1-22-10-7-17(25)13-16(22)4-5-20-19(22)8-11-23(2)18(9-12-24(20,23)27)15-3-6-21(26)28-14-15/h3,6,14,16-20,25,27H,4-5,7-13H2,1-2H3/t16-,17+,18-,19+,20-,22+,23-,24+/m1/s1

InChI key

QEEBRPGZBVVINN-BMPKRDENSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bufalin is a C-24 steroid with an α-pyrone ring positioned at C-17.[1] It is present in the venom glands and skin of toad, Venenum Bufonis.[2] Bufalin is a cardiotonic steroid[3] and is digoxin-like component of Chan Su.[4]

Application

Bufalin has been used:
  • to test its effect on invasion and activation of nuclear factor kappa- B (NF-κB) cells and tumor necrosis factor-α (TNF-α) stimulated rheumatoid arthritis fibroblast-like synoviocytes (RAFLSs)[2]
  • as a nuclear receptor coactivator 3 (NCOA3) inhibitor in chondrocytes[5]
  • to test its anti-metastasis effect in lung cancer cells (NCI-H460) resistant to gefitinib[4]

Biochem/physiol Actions

Bufalin is an anti-metastasis drug[4], which elicits anti-tumor functionality by favoring cell cycle arrest and apoptosis in various cancer types including hepatocellular, bladder and osteosarcoma.[2] It also promotes autophagic cell death and possesses anti-inflammatory properties.[3] Bufalin is a steroid receptor coactivator (SRC) inhibitor.[6]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Dong-Ze Qiu et al.
BMC complementary and alternative medicine, 13, 185-185 (2013-07-23)
Hepatocellular carcinoma (HCC) is a common and aggressive cancer, and the treatment options are limited for patients with advanced HCC. Bufalin, the major digoxin-like component of the traditional Chinese medicine Chansu, exhibits significant anti-tumor activities in many tumor cell lines.
Peihao Yin et al.
International journal of nanomedicine, 7, 3961-3969 (2012-08-14)
Recent studies have shown that bufalin has a good antitumor effect but has high toxicity, poor water solubility, a short half-life, a narrow therapeutic window, and a toxic dose that is close to the therapeutic dose, which all limit its
Yanlan Li et al.
Anti-cancer drugs, 30(7), e0770-e0770 (2019-03-05)
Breast cancer causes high mortality among females worldwide. Bufalin has recently been shown to trigger tumor cell death, although the mechanism of cytotoxicity remains unclear. The cytotoxicity of bufalin in breast cancer cells was examined using an MTT assay. The
Qingfeng Meng et al.
American journal of translational research, 8(11), 4968-4974 (2016-12-03)
Cinobufacini (Huachansu) is a Chinese medicine prepared from the skin of Bufo bufo gargarizans Cantor (Bufonidae), and has long been used in traditional Chinese medicine. In the present study, the anti-retinoblastoma constituent bufalin obtained from Cinobufacini was investigated. Treatment of
Lingling Tong et al.
Cancer medicine, 8(10), 4782-4791 (2019-06-30)
In the present study, we evaluated the expression and function of human long noncoding RNA (lncRNA) activated by DNA damage (NORAD) in human epithelial ovarian cancer (EOC). NORAD expression was evaluated by qRT-PCR in EOC cell lines and in situ

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service