AXYTR222YRS
Pipette reference tips
racked, size 200 μL, pkg of 5x960tips/cs, yellow, sterile
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
yellow
sterility
sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter: no
graduation
manufacturer/tradename
Axygen Inc TR-222-Y-R-S
Corning
packaging
pkg of 5x960tips/cs
size
200 μL
color
yellow
Looking for similar products? Visit Product Comparison Guide
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service