Skip to Content
Merck
All Photos(1)

Key Documents

AXYQTF1100CBKRS

Pipette filter tips for Qiagen®

capacity 1000 μL, black, conductive, racked, pkg of 16x96tips/cs, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
capacity:
1000 μL
sterility:
sterile
packaging:
pkg of 16x96tips/cs
manufacturer/tradename:
Corning QTF-1100-CBK-R-S
Pricing and availability is not currently available.

sterility

sterile

manufacturer/tradename

Corning QTF-1100-CBK-R-S

packaging

pkg of 16x96tips/cs

capacity

1000 μL

color

black

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Axygen® 96-well tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
  • Clear and conductive black polypropylene tips are available in 96-well format
  • Certified RNase-/DNase-free, nonpyrogenic
  • Sterile tips for sensitive biological applications
  • Aerosol resistant filtered tips reduce carryover contamination

Other Notes

Browse all pipette tips

Legal Information

Axygen is a registered trademark of Corning, Inc.
Qiagen is a registered trademark of Qiagen Group

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service