AXYPCR05LC
MAXYMum Recovery® PCR tubes
with flat caps, thin wall, size 0.5 mL, pkg of 10x1000tubes/cs
Synonym(s):
centrifuge tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
Recommended Products
material
cap (snap cap)
conical bottom tube
polypropylene
polypropylene cap
sterility
non-sterile
feature
graduations: no
manufacturer/tradename
Axygen Inc PCR-05-L-C
Corning
packaging
pkg of 10x1000tubes/cs
size
0.5 mL
color
clear
fitting
cap (push fit)
suitability
suitable for PCR
binding type
(ultra-low retention)
Looking for similar products? Visit Product Comparison Guide
Legal Information
Maxymum Recovery is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service