Skip to Content
Merck

AV44019

Anti-SLC30A1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Solute carrier family 30 (Zinc transporter), member 1, Anti-ZNT1, Anti-ZRC1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC30A1(7779)

General description

Solute carrier family 30, member 1 (SLC30A1, ZNT1, ZRC1) is a zinc transporter involved in regulating zinc and other divalent cation flux. SLC30A1/ ZNT1 is frequently coexpressed with L-type voltage-dependent calcium channel (LTCC), a major route for zinc influx. SLC30A1/ ZNT1 is believed to regulate cellular ion (calcium, cadmium, zinc) homeostasis, at least in part, by modulating/inhibiting L-type calcium channels.

Immunogen

Synthetic peptide directed towards the middle region of human SLC30A1

Application

Anti-SLC30A1 polyclonal antibody is used to tag solute carrier family 30, member 1 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of solute carrier family 30, member 1 in divalent cation (Zn2+, Cd2+, Ca2+) homeostasis, especially in conjunction with L-type voltage-dependent calcium channels.

Biochem/physiol Actions

Anti-SLC30A1 polyclonal antibody reacts with human, canine, and pig solute carrier family 30, member 1/ZNT1 proteins.
SLC30A1 may be involved in zinc transport out of the cell.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: FCVNPCFPDPCKAFVEIINSTHASVYEAGPCWVLYLDPTLCVVMVCILLY

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library