Skip to Content
Merck
All Photos(1)

Key Documents

AV30213

Sigma-Aldrich

Anti-RGS4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC2124, Anti-MGC60244, Anti-RGP4, Anti-Regulator of G-protein signalling 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

23 kDa

species reactivity

mouse, rat, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RGS4(5999)

General description

RGS4 is known to regulate G protein signaling. RGS4 polymorphisms have been analysed for its linkages to schizophrenia.
Rabbit Anti-RGS4 antibody recognizes human, mouse, rat, rabbit, bovine, and canine RGS4.

Immunogen

Synthetic peptide directed towards the C terminal region of human RGS4

Application

Rabbit Anti-RGS4 antibody can be used for western blot (1.0μg/ml) applications.

Biochem/physiol Actions

Regulator of G protein signaling (RGS) family members are regulatory molecules that act as GTPase activating proteins (GAPs) for G alpha subunits of heterotrimeric G proteins. RGS proteins are able to deactivate G protein subunits of the Gi alpha, Go alpha and Gq alpha subtypes. They drive G proteins into their inactive GDP-bound forms. Regulator of G protein signaling 4 belongs to this family. All RGS proteins share a conserved 120-amino acid sequence termed the RGS domain. This protein negatively regulates signaling upstream or at the level of the heterotrimeric G protein and is localized in the cytoplasm.Regulator of G protein signaling (RGS) family members are regulatory molecules that act as GTPase activating proteins (GAPs) for G alpha subunits of heterotrimeric G proteins. RGS proteins are able to deactivate G protein subunits of the Gi alpha, Go alpha and Gq alpha subtypes. They drive G proteins into their inactive GDP-bound forms. Regulator of G protein signaling 4 belongs to this family. All RGS proteins share a conserved 120-amino acid sequence termed the RGS domain. Regulator of G protein signaling 4 protein is 37% identical to RGS1 and 97% identical to rat Rgs4. This protein negatively regulate signaling upstream or at the level of the heterotrimeric G protein and is localized in the cytoplasm.

Sequence

Synthetic peptide located within the following region: EAQKKIFNLMEKDSYRRFLKSRFYLDLVNPSSCGAEKQKGAKSSADCASL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Zeng et al.
The Journal of biological chemistry, 273(52), 34687-34690 (1998-12-18)
Regulators of heterotrimeric G protein signaling (RGS) proteins are GTPase-activating proteins (GAPs) that accelerate GTP hydrolysis by Gq and Gi alpha subunits, thus attenuating signaling. Mechanisms that provide more precise regulatory specificity have been elusive. We report here that an
Xin Lan et al.
Nature communications, 15(1), 6350-6350 (2024-07-28)
The arginyl-transferase ATE1 is a tRNA-dependent enzyme that covalently attaches an arginine molecule to a protein substrate. Conserved from yeast to humans, ATE1 deficiency in mice correlates with defects in cardiovascular development and angiogenesis and results in embryonic lethality, while
Kodavali V Chowdari et al.
Human molecular genetics, 11(12), 1373-1380 (2002-05-25)
Gene expression analyses of postmortem cerebral cortex suggest that transcription of the regulator of G-protein signaling 4 (RGS4) is decreased in a diagnosis-specific manner in subjects with schizophrenia. To evaluate the possible role of RGS4 in the pathogenesis of schizophrenia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service