Skip to Content
Merck
All Photos(1)

Key Documents

AV100609

Sigma-Aldrich

Anti-GLI1 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Glioma-associated oncogene homolog 1 (zinc finger protein)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

118 kDa

species reactivity

human, rabbit, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GLI1(2735)

Related Categories

Immunogen

Synthetic peptide directed towards the N terminal region of human GLI1

Application

Anti-GLI1 (AB2) antibody produced in rabbit is suitable for western blotting at a concentration of 1.25 μg/ml.

Biochem/physiol Actions

GLI1 is a family of zinc finger transcription factors that mediate the effects of Hedgehog pathway. The activation of GLI1 is the effect of the binding of Shh ligand to receptor PTCH. GLI1 has been observed to be overexpressed in glioblastoma and induces the genes involved in tumor development and progression.

Sequence

Synthetic peptide located within the following region: LRPLPSQGAPSVGTEGLSGPPFCHQANLMSGPHSYGPARETNSCTEGPLF

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard L Carpenter et al.
Discovery medicine, 13(69), 105-113 (2012-03-01)
The Hedgehog signaling pathway regulates normal cell growth and differentiation. When deregulated, the Hedgehog pathway leads to tumorigenesis and supports more aggressive phenotypes of human cancers, such as progression, metastasis, and therapeutic resistance. The glioma-associated oncogene homolog 1 (GLI1) family
Hu Zhu et al.
Current genomics, 11(4), 238-245 (2010-12-02)
Sonic hedgehog (Shh) signaling is critically important for embryogenesis and other cellular processes in which GLI transcription factors mediate the terminal effects of the pathway. GLI1, in particular, plays a significant role in human cancers. Consequently, GLI1 and its upstream

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service