Skip to Content
Merck
All Photos(1)

Key Documents

ABK0005

Sigma-Aldrich

HumiBridge ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

antibody form

IgG fraction of antiserum

species reactivity

human (Human IgG)

shipped in

wet ice

storage temp.

2-8°C

General description

A Protein Conformational Array (PCA) in a sandwich ELISA format where the plate is coated with a panel of antibodies raised against peptides derived from the full length protein sequence of Adalimumab. Taken individually, each of these antibodies is strongly antigenic to the peptide sequence that was used in its production. However, when these peptides are incorporated into a full length correctly folded protein, the antigenicity of many of them is masked by the three dimensional structure of the protein and only a limited number of the antibodies respond. The result is a histogram which can be likened to a ‘fingerprint′ for correctly folded Adalimumab. For a Humira Biosimilar, if the protein is correctly folded and glycosylated, the ‘fingerprint′ will match that of Adalimumab. If it is not correctly folded, previously masked peptide sequences will be exposed and will be recognized by the antibody made to that exposed sequence. In this way, changes in the ‘fingerprint′ generated by the ELISA will point out differences between the Biosimilar and authentic Adalimumab.

Application

For research use only. Not for use in diagnostic procedures.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanli Song et al.
mAbs, 10(3), 397-405 (2018-01-10)
The elucidation of antibody higher order structure (HOS) is critical in therapeutic antibody development. Since HOS determines the protein bioactivity and chemo-physical properties, this knowledge can help to ensure that the safety and efficacy attributes are not compromised. Protein conformational
Biosimilar mAb in-process Sample Higher Order Structure Analysis with Protein Conformational Array ELISA.
Michael Davies, Gan Wang, Jian Gong, Guofeng Fu and Xing Wang
Pharmaceut. Res., 1-11 (2016)
Xing Wang et al.
Frontiers in pharmacology, 4, 103-103 (2013-08-24)
Antibody arrays were developed to probe a monoclonal antibody's three-dimensional structure (3-D structure). Peptides with overlapping regions were designed to cover the whole mAb light chain and heavy chain, respectively, and used to generate polyclonal antibodies after the conjugation of
Soon Kwan Jung et al.
mAbs, 6(5), 1163-1177 (2014-12-18)
Remsima (infliximab) was recently approved as the world's first biosimilar monoclonal antibody (mAb) in both the European Union and Korea. To achieve this, extensive physicochemical characterization of Remsima in relation to Remicade was conducted in order to demonstrate the highly

Articles

Monoclonal antibodies (mAbs) lead biologics growth, predicted $125 billion worldwide sales by 2020, emphasizing their therapeutic significance.

Monoclonal antibodies (mAbs) lead biologics growth, predicted $125 billion worldwide sales by 2020, emphasizing their therapeutic significance.

Monoclonal antibodies (mAbs) lead biologics growth, predicted $125 billion worldwide sales by 2020, emphasizing their therapeutic significance.

Monoclonal antibodies (mAbs) lead biologics growth, predicted $125 billion worldwide sales by 2020, emphasizing their therapeutic significance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service