Aldehyde dehydrogenase from bakers yeast, product A9770, is a tetramer consisting of subunits of 57 kDa. The molecular weights of its monomer, dimer, trimer, and tetramer are 57 kDa, 114 kDa, 171 kDa, and 228 kDa respectively. The monomers are thought to be assembled into tetramers in a heterologous square arrangement.
A9770
Aldehyde Dehydrogenase, potassium-activated from yeast
lyophilized powder, ≥10 units/mg protein
Synonym(s):
ALDH, CoA-independent aldehyde dehydrogenase, m-methylbenzaldehyde dehydrogenase, Aldehyde:NAD[P]+ oxidoreductase
About This Item
Recommended Products
biological source
yeast
form
lyophilized powder
specific activity
≥10 units/mg protein
storage condition
(Tightly closed. Dry)
color
white
UniProt accession no.
foreign activity
NADH oxidase ≤0.01%
NADPH oxidase ≤0.01%
alcohol dehydrogenase ≤0.01%
lactic dehydrogenase ≤0.01%
storage temp.
2-8°C
General description
Research area: Cell Signaling
Application
Biochem/physiol Actions
Unit Definition
Physical form
antibody
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.
Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.
Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.
Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.
-
What is the molecular weight of Aldehyde Dehydrogenase, potassium-activated from baker's yeast (S. cerevisiae), product A9770?
1 answer-
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service