Skip to Content
Merck
All Photos(1)

Key Documents

A9688

Sigma-Aldrich

Anti-Mouse IgM (μ-chain specific)–Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Goat Anti-Mouse IgM (μ-chain specific)–AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

mouse

technique(s)

direct ELISA: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50
western blot: 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Binds mouse IgM; does not bind other mouse Igs.
IgM is a glycoprotein antibody that regulates humoral immune responses. Mouse IgM is involved in modulating B cell memory. This antibody isotype has also been implicated in the development of autoimmune responses associated with the pathogenesis of type 1 diabetes in mice.
Goat Anti-Mouse IgM (μ-chain specific)-Alkaline Phosphatase antibody is specific for mouse IgM when tested against purified mouse IgA, IgG and IgM myeloma proteins.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Enzyme-linked immunosorbent assay (1 paper)
Goat Anti-Mouse IgM (μ-chain specific)-Alkaline Phosphatase antibody has been used for ELISA and immunofluorescence assays. The antibody can also be used for IHC (1:50) and western blot (1:30,000) applications.
Mouse plasma antibody isotype determination was performed by ELISA using alkaline phopshatase-conjugated goat anti-mouse IgM as the secondary antibody.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1 mM MgCl2, 10 mM glycine, 1% bovine serum albumin, 50% glycerol and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadia Malagolini et al.
Glycobiology, 17(7), 688-697 (2007-03-31)
The carbohydrate determinants Sd(a) and sialyl Lewis x (sLex) both result from substitution of an alpha2,3-sialylated type 2 chain: the first with an N-acetylgalactosamine (GalNAc) beta1,4-linked to Gal and the second by an alpha1,3-linked fucose on N-acetylglucosamine. The Sd(a) antigen
Sarah N Lauder et al.
Wellcome open research, 2, 1-1 (2017-02-28)
Background. The myeloid enzyme 12/15-lipoxygenase (LOX), which generates bioactive oxidized lipids, has been implicated in numerous inflammatory diseases, with several studies demonstrating an improvement in pathology in mice lacking the enzyme. However, the ability of 12/15-LOX to directly regulate B
Óscar Mariño-Crespo et al.
Oncology letters, 15(1), 580-587 (2018-02-03)
The CDw75 epitope is an α(2,6) sialylated antigen overexpressed in colorectal cancer (CRC), where its expression correlates with the progression of the disease. The CDw75 epitope is located mainly in N-glycoproteins, whose identity remains unknown. The aim of the present
M P Lunn et al.
Journal of neurochemistry, 75(1), 404-412 (2000-06-15)
Gangliosides, sialic acid-bearing glycosphingolipids, are highly enriched in the vertebrate nervous system. Anti-ganglioside antibodies are associated with various human neuropathies, although the pathogenicity of these antibodies remains unproven. Testing the pathogenic role of anti-ganglioside antibodies will be facilitated by developing
Dimitrios Tsiantoulas et al.
Scientific reports, 7(1), 3540-3540 (2017-06-16)
Mice lacking secreted IgM (sIgM -/-) antibodies display abnormal splenic B cell development, which results in increased marginal zone and decreased follicular B cell numbers. However, the mechanism by which sIgM exhibit this effect is unknown. Here, we demonstrate that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service