Skip to Content
Merck

Skip To

A9228

Amyloglucosidase from Rhizopus sp.

≥40,000 units/g solid

Synonym(s):

1,4-α-D-Glucan glucohydrolase, Exo-1,4-α-glucosidase, Glucoamylase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-877-2
MDL number:
EC Number:
Specific activity:
≥40,000 units/g solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized solid

Quality Segment

specific activity

≥40,000 units/g solid

solubility

0.03 M sodium citrate-phosphate buffer, pH 4.5: soluble 5.0 mg/mL

storage temp.

−20°C

Application

Amyloglucosidase is used to hydrolyze α-D-glucosides. It may be used in the brewing of beer and in the production of bread and juices. Amyloglucosidase, from Rhizopus sp., has been used to study the cleavage of oligosaccharides during ER-associated degradation of proteins (ERAD).[1] The enzyme has been used in the glycosylation of N-vanillyl-nonanamide to form a water-soluble component with pharmacological applications.[2] The glycogen was estimated in stipe residue of Coprinus cinereus by treating with amyloglucosidase and measuring the amount of glucose produced.

Biochem/physiol Actions

Amyloglucosidases are capable of hydrolyzing the α-D-(1-4), the α-D-(1-6), and the α-D-(1-3) glucosidic bonds of oligosaccharides [3].

Physical form

Lyophilized salt free powder

Other Notes

One unit will liberate 1.0 mg of glucose from starch in 3 min at pH 4.5 at 55 °C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A16021011310115
specific activity

≥40,000 units/g solid

specific activity

≥40 units/mg protein

specific activity

~120 U/mg

specific activity

~70 U/mg

form

lyophilized solid

form

ammonium sulfate suspension

form

powder

form

powder

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

0.03 M sodium citrate-phosphate buffer, pH 4.5: soluble 5.0 mg/mL

solubility

-

solubility

-

solubility

-

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters