Skip to Content
Merck
All Photos(2)

Key Documents

A8452

Sigma-Aldrich

Monoclonal Anti-α-Fetoprotein (AFP) antibody produced in mouse

ascites fluid, clone C3

Synonym(s):

Afp Antibody, Afp Antibody - Monoclonal Anti-α-Fetoprotein (AFP) antibody produced in mouse, Alpha Fetoprotein Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

C3, monoclonal

contains

15 mM sodium azide

species reactivity

pig, canine, human

should not react with

mouse, cat, rat, bovine

technique(s)

dot blot: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:500 using human fetal liver
indirect ELISA: suitable

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AFP(174)

General description

α-Fetoprotein (AFP) is a one of the main glycoprotein present in the fetal liver, gastrointestinal tract, and yolk sac. AFP levels in amniotic fluid have been used as biomarkers for anencephaly and spina bifida.
Monoclonal Anti-α-Fetoprotein (AFP) (mouse IgG2a isotype) is derived from the C3 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with purified human α-fetoprotein.

Immunogen

human α-fetoprotein

Application

Monoclonal Anti-α-Fetoprotein (AFP) antibody produced in mouse has been used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • dot blot
  • immunocytochemistry
  • immunofluorescence
  • immunocytochemistry

Mouse Monoclonal Anti-α-Fetoprotein has been used for immunocytochemistry. The product can also be used for dot blot and indirect ELISA.

Biochem/physiol Actions

Increased concentrations of α-fetoprotein (AFP) in serum are less common in patients with malignancies of the gastrointestinal tract and of other organ systems with massive hepatic metastases. Hence, AFP determination is useful for the diagnosis of certain germ line tumors that contain yolk sac structures (testicular and ovarian tumors), hepatocellular carcinoma (hepatoma), and some malignant tumors of the gastrointestinal tract. AFP serves as an important marker for certain genetic and embryonic defects. Routine screening of pregnancies with a risk of neural tube defects (spina bifida, anencephaly, and Down′s syndrome) may be facilitated by the monitoring of AFP levels.

Target description

AFP is a plasma protein found highly expressed in the human fetus where it functions to block the transport of estradiol across the placenta.

Physical form

The product is provided as ascites fluid with 0.1% sodium azide as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alpha-fetoprotein in ontogenesis and its association with malignant tumors.
G I Abelev
Advances in cancer research, 14, 295-358 (1971-01-01)
Alpha-fetoprotein in the antenatal diagnosis of anencephaly and spina bifida.
D J Brock et al.
Lancet (London, England), 2(7770), 197-199 (1972-07-29)
Reduced PLP1 expression in induced pluripotent stem cells derived from a Pelizaeus-Merzbacher disease patient with a partial PLP1 duplication
Shimojima K, et al.
Journal of Human Genetics, 57(9), 580-580 (2012)
Characterization of a novel embryonic stem cell line from an ICSI-derived blastocyst in the African green monkey
Shimozawa N, et al.
Reproduction (Cambridge, England), 139(3), 565-565 (2010)
Mark O Clements et al.
Tissue engineering, 12(7), 1741-1751 (2006-08-08)
Human stem cells could revolutionize the field of medicine by providing a diverse range of cell types for tissue replacement therapies and drug discovery. To achieve this goal, genetic tools need to be optimized and developed for controlling and manipulating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service