Skip to Content
Merck

A7772

S-(5′-Adenosyl)-L-cysteine

>98% (TLC), suitable for ligand binding assays

Synonym(s):

S-(5′-deoxyadenosin-5′-yl)-L-cysteine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H18N6O5S
CAS Number:
Molecular Weight:
370.38
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

S-(5′-Adenosyl)-L-cysteine,

assay

>98% (TLC)

Quality Segment

form

powder

technique(s)

ligand binding assay: suitable

storage temp.

−20°C

SMILES string

NC(CSCC1OC(C(O)C1O)n2cnc3c(N)ncnc23)C(O)=O

InChI

1S/C13H18N6O5S/c14-5(13(22)23)1-25-2-6-8(20)9(21)12(24-6)19-4-18-7-10(15)16-3-17-11(7)19/h3-6,8-9,12,20-21H,1-2,14H2,(H,22,23)(H2,15,16,17)

InChI key

RVFHZLGRQFCOKV-UHFFFAOYSA-N

Biochem/physiol Actions

S-adenosyl-L-cysteine, an analogue of S-adenosyl-L-homocysteine (L-AdoHcy), is used in the cyrstallizaton of methytransferases such as XMT and DXMT N-methyltransferases.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library