Skip to Content

A7075

Adrenocorticotropic Hormone rat

≥95% (HPLC)

Synonym(s):

Adrenocorticotropic hormone fragment 1-39, Corticotropin A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C210H315N57O57S
CAS Number:
Molecular Weight:
4582.16
UNSPSC Code:
51111800
NACRES:
NA.32
MDL number:
Biological source:
rat
Form:
powder
Assay:
≥95% (HPLC)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

sterility

non-sterile

assay

≥95% (HPLC)

form

powder

solubility

1% acetic acid: 1.00-1.04 mg/mL, clear, colorless, water: 1.00-1.04 mg/mL, clear, colorless

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

rat ... Pomc(24664)

Biochem/physiol Actions

Stimulates synthesis and secretion of glucocorticoids by adrenal cortex.

Other Notes

Lyophilized from 0.1% TFA in H2O


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library