Skip to Content
Merck
All Photos(1)

Key Documents

A7064

Sigma-Aldrich

Anti-Biotin−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Biotin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

technique(s)

direct ELISA: 1:30,000
dot blot: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Biotin is a carboxylase co-enzyme that regulates the metabolism of amino acids, glucose and fatty acids. This co-enzyme has been implicated in processes such as immune responses, cell division and fetal growth. Biotin may also be involved in the epigenetic modifications of chromatin.
Goat Anti-Biotin-Alkaline Phosphatase antibody is specific for biotin when tested against biotin-goat IgG, goat IgG, and KLH.
Specificity and reactivity are determined by Ouchterlony Double Diffusion and immunoelectrophoresis versus the biotin-carrier protein complex. No reactivity is observed versus the carrier protein.

Immunogen

Biotin-KLH conjugate

Application

ELISA Assays using human spleen cells were performed using alkaline phosphatase-conjugated goat anti-biotin as the secondary antibody with incubation for 1.5 hours at room temperature. Purified IgG from K/BxN, BxSB or C57BL/6 mouse serum was analyzed by western blot using alkaline phosphatase-conjugated goat anti-biotin antibody as the secondary.
Goat Anti-Biotin-Alkaline Phosphatase antibody has been used for dot blot assays. The product can also be used for ELISA (1:30,000) and immunohistochemistry (1:50).
In some applications, localization of biotinylated probes with avidin produces high background levels. Anti-biotin reagents may be substituted for avidin to decrease non-specific binding.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 10 mM glycine, 1 mM MgCl2, 1% bovine serum albumin, 50% glycerol, and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heike Albert et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(39), 15005-15009 (2008-09-26)
IgG antibodies are potent inducers of proinflammatory responses. During autoimmune diseases such as arthritis and systemic lupus erythematosus, IgG autoantibodies are responsible for the chronic inflammation and destruction of healthy tissues by cross-linking Fc receptors on innate immune effector cells.
Peter A Suci et al.
Langmuir : the ACS journal of surfaces and colloids, 22(21), 8891-8896 (2006-10-04)
Protein cage architectures such as viral capsids, heat shock proteins, ferritins, and DNA-binding proteins are nanoscale modular subunits that can be used to expand the structural and functional range of composite materials. Here, layer-by-layer (LbL) assembly was used to incorporate
Peter A Suci et al.
Chemistry & biology, 14(4), 387-398 (2007-04-28)
Nanomedicine directed at diagnosis and treatment of infections can benefit from innovations that have substantially increased the variety of available multifunctional nanoplatforms. Here, we targeted a spherical, icosahedral viral nanoplatform to a pathogenic, biofilm-forming bacterium, Staphylococcus aureus. Density of binding
Taiji Nozaki et al.
Journal of immunology (Baltimore, Md. : 1950), 179(8), 5238-5245 (2007-10-04)
Rejected MHC-mismatched cardiac allografts in CCR5(-/-) recipients have low T cell infiltration, but intense deposition of C3d in the large vessels and capillaries of the graft, characteristics of Ab-mediated rejection. The roles of donor-specific Ab and CD4 and CD8 T
Rocio Rodriguez-Melendez et al.
The Journal of nutritional biochemistry, 14(12), 680-690 (2003-12-24)
In mammals, biotin serves as coenzyme for four carboxylases, which play essential roles in the metabolism of glucose, amino acids, and fatty acids. Biotin deficiency causes decreased rates of cell proliferation, impaired immune function, and abnormal fetal development. Evidence is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service