Skip to Content
Merck
All Photos(1)

Key Documents

A7058

Sigma-Aldrich

Anti-polyHistidine−Peroxidase antibody, Mouse monoclonal

clone HIS-1, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-polyHistidine, Monoclonal 6 His epitope tag, Monoclonal 6xHis-tag, Monoclonal HHHHHH epitope tag, Monoclonal Hexa His tag, Monoclonal His-tag, Monoclonal His6 tag, Monoclonal Histidine tagged, Monoclonal Poly-His-tag

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.56
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

HIS-1, monoclonal

form

lyophilized powder

packaging

vial of 0.5 mL

concentration

5-11 mg/mL

technique(s)

western blot: 1:2,000 using lysates of Escherichia coli induced to express a 6xHis tagged protein

isotype

IgG2a

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

The monoclonal anti-polyHistidine Peroxidase Conjugate antibody recognizes native and denatured forms of synthetic polyhistidine or polyhistidine-tagged fusion proteins. The product is reactive with fusion proteins expressed by prokaryotic pET, pRSET and pTrc expression vectors.

Specificity

The antibody recognizes synthetic polyHistidine, as well as native or denatured, reduced forms of proteins tagged with 6X histidines, expressed in selected vectors.

Immunogen

recombinant polyHistidine tagged fusion protein.

Application

Antibody suitable for immunoblotting. Working dilution 1:20



Also suitable for dot blot assays and ELISA

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% MIT.

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Reconstitution

The antibody conjugate should be reconstituted with 0.5 ml deionized water.

Legal Information

This Product is covered by patent DE 19507166 and foreign equivalents exclusively licensed to QIAGEN GmbH, Qiagen Strasse 1, D-40724 Hilden, Germany.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Zhang et al.
Nature communications, 6, 8635-8635 (2015-10-27)
Phenylpropanoids comprise an important class of plant secondary metabolites. A number of transcription factors have been used to upregulate-specific branches of phenylpropanoid metabolism, but by far the most effective has been the fruit-specific expression of AtMYB12 in tomato, which resulted
Renhua Huang et al.
PloS one, 11(1), e0145872-e0145872 (2016-01-06)
Affinity reagents of high affinity and specificity are very useful for studying the subcellular locations and quantities of individual proteins. To generate high-quality affinity reagents for human Lyn tyrosine kinase, a phage display library of fibronectin type III (FN3) monobodies
Laurence Guglielmi et al.
Methods in molecular biology (Clifton, N.J.), 562, 215-224 (2009-06-26)
The most frequently used approach to produce single-chain Fv fragments (scFv) and Fab in Escherichia coli is to express them in the periplasm of the bacteria. We present here an alternative procedure that uses cytoplasmic expression of soluble active scFv.
Rita Figueira et al.
Scientific reports, 5, 17524-17524 (2015-12-02)
The Escherichia coli eukaryote-like serine/threonine kinase, encoded by yeaG, is expressed in response to diverse stresses, including nitrogen (N) starvation. A role for yeaG in bacterial stress response is unknown. Here we reveal for the first time that wild-type E.
Philippe Thullier et al.
PloS one, 8(5), e65855-e65855 (2013-06-07)
The lethal toxin (LT) of Bacillus anthracis, composed of the protective antigen (PA) and the lethal factor (LF), plays an essential role in anthrax pathogenesis. PA also interacts with the edema factor (EF, 20% identity with LF) to form the

Questions

  1. To whom it may concern. Is the antibody Anti-his 6 tag hrseradish Peroxidase conjugated? Thank you

    1 answer
    1. This product is horseradish peroxidase conjugated.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service