Skip to Content
Merck

A6929

ACCUSPIN System-Histopaque®-1077

sterile-filtered, density: 1.077 g/mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

sterility

aseptically filled

form

solution

shelf life

Expiry date on the label.

packaging

pkg of packaging 40 tubes

technique(s)

cell culture | mammalian: suitable

density

1.077 g/mL

application(s)

hematology
histology

storage temp.

2-8°C

General description

ACCUSPIN System-Histopaque-1077 products are intended for use in the isolation of lymphocytes and other mononuclear cells. The separation medium, Histopaque-1077, is a sterile-filtered, endotoxin-tested solution of polysucrose and sodium diatrizoate, adjusted to a density of 1.077 g/mL. ACCUSPIN System-HISTOPAQUE-1077 employs γ-irradiated 12mL polypropylene centrifuge tubes specially designed with two chambers separated by a porous high-density polyethylene barrier ("frit"). Each tube is aseptically filled with 3 mL of Histopaque 1077. Each tube will separate 3-6 mL of anticoagulated blood.

Application

ACCUSPIN System-HISTOPAQUE-1077 employs centrifuge tubes specially designed with two chambers separated by a porous high-density polyethylene barrier ("frit"). The lower chamber contains HISTOPAQUE-1077 which allows the addition of anticoagulated whole blood without risk of mixing with the separation medium. On centrifugation, the whole blood descends through the frit to contact with the HISTOPAQUE-1077 below the frit, giving a clear separation of the blood components. The erythrocytes aggregate and the granulocytes become slightly hypertonic, increasing their sedimentation rate, resulting in pelleting at the bottom of the ACCUSPIN tube. Lymphocytes and other mononuclear cells, i.e., monocytes, remain at the plasma-HISTOPAQUE-1077 interface.
ACCUSPIN System-Histopaque-1077 is used for the separation of lymphocytes and other mononuclear cells from whole blood and bone marrow using density gradient separation media. Histopaque-1077 has been used for:

  • human lymphocyte antigen (HLA) typing
  • initial isolation step prior to the enumeration of T, B, and ‘null’ lymphocytes.
  • preparation of pure lymphocyte suspensions for cell culture and cytotoxicity assays.
  • isolation and purification of peripheral blood mononuclear cells (PBMCs) and bone marrow mononuclear cells (BMMCs)citation
  • isolation of human lymphocytescitation
  • isolation of breast cancer circulating tumor cells (CTCs) from the whole bloodcitation

Biochem/physiol Actions

The lower chamber contains HISTOPAQUE-1077 which allows the addition of anticoagulated whole blood without risk of mixing with the separation medium. On centrifugation, the whole blood descends through the frit to contact with the HISTOPAQUE-1077 below the frit, giving a clear separation of the blood components. The erythrocytes aggregate and the granulocytes become slightly hypertonic, increasing their sedimentation rate, and resulting in pelleting at the bottom of the ACCUSPIN tube. Lymphocytes and other mononuclear cells, i.e., monocytes, remain at the plasma-HISTOPAQUE-1077 interface.

Features and Benefits

ACCUSPIN System-Histopaque®-1077 offers numerous advantages, such as:
  • Optimal recovery of viable cells
  • Permits selective separation of blood cell types
  • Prevents cell distortion
  • Preserves cell viability
  • Minimal extraneous cell interference
  • Reproducible performance from batch to batch
  • Histopaque is sterile-filtered and endotoxin-tested
  • Stable at 2-8°C for at least 2 years when protected from light

Preparation Note

Package contains 40 prefilled tubes

Legal Information

Accuspin is a trademark of Sigma-Aldrich Co. LLC
Histopaque is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Tiffany A Greenwood et al.
PloS one, 7(1), e29630-e29630 (2012-01-19)
While it is clear that schizophrenia is highly heritable, the genetic basis of this heritability is complex. Human genetic, brain imaging, and model organism studies have met with only modest gains. A complementary research tactic is to evaluate the genetic
Nicolas Antoine-Moussiaux et al.
PloS one, 4(5), e5728-e5728 (2009-05-30)
In a recent study dealing with a mouse model of Trypanosoma evansi-associated disease, a remarkable synchrony between the parasitaemia peak and the white-blood-cell count nadir was noticed. The present study was designed to establish whether there is a direct causal



Global Trade Item Number

SKUGTIN
A6929-40X3ML04061833383070