Skip to Content
Merck
All Photos(1)

Key Documents

A6917

Sigma-Aldrich

Anti-Horse IgG (whole molecule)−Peroxidase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Rabbit Anti-Horse IgG (whole molecule)−HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:10,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

An immunoglobulin has two heavy chain and two light chain connected by disulfide bond. It mainly helps in immune defense. It is a glycoprotein. IgG is a major class of immunoglobulin. Horse IgG has different subclasses, IgG2a, IgG2b, IgG2c, IgG1 and IgG(T). Compared to other IgG molecules, IgG(T) has more carbohydrate content. In healthy individuals, total IgG is dispersed equally between circulating plasma and interstitial fluids.
Horse IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in horse serum. IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection.

Immunogen

Purified horse IgG

Application

Anti-Horse IgG (whole molecule)-Peroxidase antibody produced in rabbit has been used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • HeV soluble G
  • indirect ELISA (HeVsG iELISA)

Biochem/physiol Actions

Equine IgG antibodies mainly regulate mucosal and systemic immunological responses and thereby, provide protection against disease-causing pathogens such as Streptococcus equi, and the horse flu virus. Horse IgG may also function to control the advancement of EHV-1 infection .
Immunoglobulin G (IgG) participates in hypersensitivity type II and type III.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4 containing 1% bovine serum albumin and 0.05% MIT

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natacha Ferreira de Oliveira et al.
Toxins, 14(10) (2022-10-27)
Viperidae snakes are the most important agents of snakebites in Brazil. The protein composition of snake venoms has been frequently analyzed by means of electrophoretic techniques, but the interaction of proteins in venoms has barely been addressed. An electrophoretic technique
Eliane Cristina de Freitas et al.
Microbiology (Reading, England), 165(3), 355-365 (2019-01-29)
Upstream open reading frames (ORFs) are frequently found in the 5'-flanking regions of genes and may have a regulatory role in gene expression. A small ORF (named cohL here) was identified upstream from the copAB copper operon in Xanthomonascitri subsp.
Giordanni C Dantas et al.
Brazilian journal of microbiology : [publication of the Brazilian Society for Microbiology], 47(2), 518-526 (2016-03-19)
Citrus canker, caused by the Gram-negative bacterium Xanthomonas citri subsp. citri (Xac), is one of the most devastating diseases to affect citrus crops. There is no treatment for citrus canker; effective control against the spread of Xac is usually achieved
Luis Eduardo R Cunha et al.
iScience, 24(11), 103315-103315 (2021-11-02)
We used the recombinant trimeric spike (S) glycoprotein in the prefusion conformation to immunize horses for the production of hyperimmune globulins against SARS-CoV-2. Serum antibody titers measured by ELISA were above 1:106, and the neutralizing antibody titer against authentic virus
Can equids be a reservoir of Leishmania braziliensis in endemic areas
Truppel JH, et al.
PLoS ONE, 9(4), e93731-e93731 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service