Skip to Content
Merck

Skip To

A6782

Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in sheep

affinity isolated antibody, buffered aqueous solution

Synonym(s):

SAM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
peroxidase conjugate
Clone:
polyclonal
Application:
ELISA (d)
Citations:
46
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

sheep

Quality Segment

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:10,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Mouse IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in mouse serum. IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection.
Horseradish Peroxidase (HRP) is an enzyme that catalyzes the conversion of chromogenic substrates such as o-phenylenediamine (OPD), 4-chloro-1-naphthol 3,3′,5,5′-tetramethylbenzidine (TMB), 3,3′-Diaminobenzidine (DAB) or 2,2′-azino-bis(3-ethylbenzothiazoline-6-sulphonic acid) (ABTS); chemiluminescent substrates such as CPS-3 (enhanced luminal) and fluorogenic substrates such as Ampliflu Red into detectable chromophores, light-emitters or fluorescers, respectively.

Application

Sheep polyclonal anti-Mouse IgG (whole molecule)–Peroxidase antibody may be used to detect and quantitate the level of IgG in mouse serum and biological fluids via chromogenic, chemoluminescent or fluorogenic immunochemical or immunohistochemical techniques. It may also be used as a secondary antibody in assays that use mouse IgG as the primary antibody. Primary T cells isolated from buffy goats were analyzed by western blot using HRP-conjugated sheep anti-mouse IgG as the secondary antibody.

Biochem/physiol Actions

Sheep polyclonal anti-Mouse IgG (whole molecule)–Peroxidase antibody reacts with mouse IgG in vitro and in mouse serum and biological fluids. It does not react with human serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin with preservative.

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Other Notes

Antibody adsorbed with human serum proteins.

Legal Information

Ampliflu is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A4416A5906A8924
biological source

sheep

biological source

goat

biological source

sheep

biological source

goat

technique(s)

direct ELISA: 1:10,000

technique(s)

direct ELISA: 1:10,000

technique(s)

direct ELISA: 1:10,000

technique(s)

direct ELISA: 1:10,000

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

peroxidase conjugate

conjugate

peroxidase conjugate

conjugate

peroxidase conjugate

conjugate

peroxidase conjugate

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters